Skip to Content
Merck
All Photos(1)

Key Documents

HPA026785

Sigma-Aldrich

Anti-SLC25A39 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CGI-69, Anti-FLJ22407, Anti-solute carrier family 25, member 39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RLQSQRPSMASELMPSSRLWSLSYTKWKCLLYCNGVLEPLYLCPNGARCATWFQDPTRFTGTMDAFVKIVRHEGTRTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 25, member 39 (SLC25A39), also called as CGI-69, with 318 amino acids is encoded by a gene with 11 exons mapped to human chromosome 17. SLC family proteins are expressed abundantly in central nervous system (CNS). Members of SLC25 family share characteristic features such as tripartite structure, six transmembrane α-helices and a 3-fold repeated signature motifs. SLC25A39 or CGI-69 transcript is widely expressed in human testis and kidney.

Immunogen

solute carrier family 25, member 39 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 25, member 39 (SLC25A39) is involved in shifting molecules across the mitochondrial membrane. The encoded protein helps in activation of manganese-containing superoxide dismutase in human. SLC25A39 regulates the mitochondrial iron homeostasis and controls iron level.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71475

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of the human 2-oxoglutarate carrier lowers mitochondrial membrane potential in HEK-293 cells: contrast with the unique cold-induced mitochondrial carrier CGI-69.
YU XX
The Biochemical Journal, 353(pt2), 369-375 (2001)
Tangled up in red: intertwining of the heme and iron-sulfur cluster biogenesis pathways.
Rouault TA, Tong WH
Cell Metabolism, 10(2), 80-81 (2009)
Discovery of genes essential for heme biosynthesis through large-scale gene expression analysis.
Nilsson R
Cell Metabolism, 10(2), 119-130 (2009)
Ferdinando Palmieri
Molecular aspects of medicine, 34(2-3), 465-484 (2012-12-26)
SLC25 is a large family of nuclear-encoded transporters embedded in the inner mitochondrial membrane and in a few cases other organelle membranes. The members of this superfamily are widespread in eukaryotes and involved in numerous metabolic pathways and cell functions.
Tatjana Haitina et al.
Genomics, 88(6), 779-790 (2006-09-05)
Members of the solute carrier family 25 (SLC25) are known to transport molecules over the mitochondrial membrane. In this paper we present 14 novel members of SLC25 family in human. These were provided with following gene symbols by the HGNC:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service