Skip to Content
Merck
All Photos(2)

Key Documents

HPA023626

Sigma-Aldrich

Anti-ITGB6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-integrin, beta 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RGDCVCGKCVCTNPGASGPTCERCPTCGDPCNSKRSCIECHLSAAGQAREECVDKCKLAGATISEEEDFSKDGSVSCSLQGENECLITFLITTDNEGKTIIHSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGB6(3694)

General description

Integrin subunit β 6 (ITGB6) is a β subunit of integrin αvβ6, which is a membrane-spanning heterodimeric glycoprotein. It is encoded by the gene mapped to human chromosome 2q24.2. ITGB6 is solely expressed on epithelial cells during embryogenesis.

Immunogen

integrin, beta 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-ITGB6 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Integrin subunit β 6 (ITGB6) contributes to colorectal cancer (CRC) pathogenesis by activating latent transforming growth factor β (TGFβ) and maintaining TGFβ-mediated epithelial-to-mesenchymal transition (EMT). Therefore, ITGB6 can be used as a serum marker for CRC. Increased expression of ITGB6 has been observed in various processes requiring tissue remodeling such as wound healing, fibrosis, and cancer. Mutation of the ITGB6 gene leads to the development of autosomal recessive amelogenesis imperfecta, alopecia, periodontitis, and emphysema. Deletion of the gene causes pulmonary dysfunction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70079

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinichi Takatsuki et al.
American journal of medical genetics. Part A, 152A(4), 1020-1025 (2010-04-02)
Owing to the large size of chromosome 2, partial monosomy of the long arm of this chromosome gives rise to many specific phenotypes. We report on a 2-month-old girl with an interstitial deletion of 2q24.2q24.3, which was confirmed by microarray-based
Amelia Meecham et al.
Gene: X, 5, 100023-100023 (2020-06-19)
Integrin αvβ6 is a membrane-spanning heterodimeric glycoprotein involved in wound healing and the pathogenesis of diseases including fibrosis and cancer. Therefore, it is of great clinical interest for us to understand the molecular mechanisms of its biology. As the limiting
Jintao Yu et al.
American journal of cancer research, 14(5), 2608-2625 (2024-06-11)
The immune escape of colon cancer and its role in the response to immunotherapies such as PD-1/PD-L1 checkpoint inhibitors have long been of great interest. The positive outcomes of immunotherapy are limited by the immunosuppressive nature of the tumor microenvironment.
Susan Bengs et al.
International journal of cancer, 145(3), 678-685 (2019-01-18)
Colorectal cancer (CRC) is one of the leading causes of cancer-related deaths worldwide and the need for novel biomarkers and therapeutic strategies to improve diagnosis and surveillance is obvious. This study aims to identify β6 -integrin (ITGB6) as a novel
Shih-Kai Wang et al.
Human molecular genetics, 23(8), 2157-2163 (2013-12-07)
Integrins are cell-surface adhesion receptors that bind to extracellular matrices (ECM) and mediate cell-ECM interactions. Some integrins are known to play critical roles in dental enamel formation. We recruited two Hispanic families with generalized hypoplastic amelogenesis imperfecta (AI). Analysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service