Skip to Content
Merck
All Photos(9)

Key Documents

HPA021851

Sigma-Aldrich

Anti-PRKCZ antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PKC2, Anti-PRKCZ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

FNRRAYCGQCSERIWGLARQGYRCINCKLLVHKRCHGLVPLTCRKHMDSVMPSQEPPVDDKNEDADLPSEETDGIAYISSSRKHDSIKDDSEDLKPVIDGMDGIKISQGLGLQDFDLIRVIGRGSYAKVLLVRLKKNDQI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCZ(5590)

General description

PRKCZ (Protein kinase C ζ) is located on chromosome 1p36.33-p36 belonging to the PKC family of serine/threonine kinases and the aPKC subfamily.

Immunogen

Protein kinase C zeta type recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRKCZ (Protein kinase C ζ) is involved in a variety of cellular processes including cell proliferation, cell motility, and cell survival. It acts as a regulatory molecule in the insulin signaling pathways. It has been suggested that PRKCZ performs downstream of phosphatidylinositol 3-kinase (PI 3-kinase) in the insulin signal pathway for the translocation of GLUT4-encoded protein. It has also been reported that PRKCZ may act as a negative regulator by phosphorylating insulin receptor substrate-1 (IRS-1), which restricts its ability to activate phosphatidylinositol 3-kinase in response to insulin. It has been documented that PRKCZ might be involved in the pathogenesis of type 2 diabetes mellitus (T2DM). PRKCZ also has been reported to be part of various signaling pathways such as ERK/MAPK activation pathway, p70 ribosomal S6 kinase signaling cascade, activation of transcription factor NF-κB. It also plays a role in the regulation of cell polarity. The study predicts that PRKCZ may have a role in ovarian cancer progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70324

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eijiro Shimada et al.
British journal of cancer, 126(9), 1289-1300 (2022-01-13)
Dedifferentiated chondrosarcoma (DDCS) is an aggressive bone tumour with a poor prognosis and no effective treatment. Because changes in DNA methylation play critical roles in DDCS, we explored the roles that DNA methylation plays in oncogenesis to potentially identify an
Yun-Feng Li et al.
World journal of gastroenterology, 9(9), 2078-2082 (2003-09-13)
To identify the susceptible gene (s) for type 2 diabetes in the previously mapped region, 1p36.33-p36.23, in Han population of North China using single nucleotide polymorphisms (SNPs) and to analyze the haplotypes of the gene (s) related to type 2
Li Zou et al.
Journal of diabetes research, 2013, 721493-721493 (2013-05-15)
Objectives. To study the correlation between the methylation of protein kinase C epsilon zeta (PRKCZ) gene promoters and type 2 diabetes mellitus (T2DM). Methods. The case-control method was implemented in 272 unrelated to one another cases in Shiyan People's Hospital.
Kelly K Y Seto et al.
PloS one, 10(4), e0123528-e0123528 (2015-04-16)
Ovarian cancer is one of the most aggressive gynaecological cancers, thus understanding the different biological pathways involved in ovarian cancer progression is important in identifying potential therapeutic targets for the disease. The aim of this study was to investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service