Skip to Content
Merck
All Photos(7)

Key Documents

HPA021147

Sigma-Aldrich

Anti-MPO antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MPO, Anti-Myeloperoxidase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:5000- 1:10000

immunogen sequence

SRDHGLPGYNAWRRFCGLPQPETVGQLGTVLRNLKLARKLMEQYGTPNNIDIWMGGVSEPLKRKGRVGPLLACIIGTQFRKLRDGDRFWWENEGVFSMQQRQALAQISLPRIICDNTGITTVSKNNIFMSNSYPRDFVNC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPO(4353)

General description

The gene MPO (myeloperoxidase) is mapped to human chromosome 17q23.1. The protein localizes in the azurophilic granules of polymorphonuclear leukocytes. It belongs to the heme peroxidase super family.

Immunogen

Myeloperoxidase Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MPO (myeloperoxidase) is responsible for generation of hypochlorous acid and anti-microbial agents by reacting with hydrogen peroxide and chloride ions. The enzymatic activity of MPO is important for lipid peroxidation and post-translational modifications in proteins, such as halogenation, nitration and oxidative crosslinking. Increase in MPO levels is associated with coronary artery disease. It is also linked with formation and rupture of cerebral aneurysms.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78212

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryosuke Hiwa et al.
Arthritis & rheumatology (Hoboken, N.J.), 69(10), 2069-2080 (2017-06-03)
Autoantibodies against myeloperoxidase (MPO) that are expressed in neutrophils play an important role in the pathogenesis of microscopic polyangiitis (MPA). We recently observed that misfolded cellular proteins are transported to the cell surface by HLA class II molecules and are
Luyao Chen et al.
Zhong nan da xue xue bao. Yi xue ban = Journal of Central South University. Medical sciences, 39(3), 217-231 (2014-04-22)
To assess the association between myeloperoxidase (MPO) gene polymorphism and coronary artery disease (CAD). Several databases were used to retrieve relevant literature up to March 2013 by keywords. A Meta-analysis was performed by Stata12.0 software to estimate the pooled odds
M Kizaki et al.
Blood, 83(7), 1935-1940 (1994-04-01)
Myeloperoxidase (MPO), present in the azurophilic granules of polymorphonuclear leukocytes, is a myeloid enzyme whose synthesis is restricted to promyelocytes. Complete hereditary MPO deficiency affects 1 in 2,000 to 4,000 individuals; however, the genetic cause of this defect is unclear.
M Brennan et al.
Journal of neuroimmunology, 112(1-2), 97-105 (2000-12-08)
EAE is a demyelinating disease which serves as an animal model for multiple sclerosis (MS). Myeloperoxidase (MPO) has been implicated in MS through its presence in invading macrophages, and by association of a -463G/A promoter polymorphism with increased risk. Also
F R DeLeo et al.
The Journal of clinical investigation, 101(12), 2900-2909 (1998-06-24)
Myeloperoxidase (MPO) deficiency is a common inherited disorder linked to increased susceptibility to infection and malignancy. We identified a novel missense mutation in the MPO gene at codon 173 whereby tyrosine is replaced with cysteine (Y173C) that is associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service