Skip to Content
Merck
All Photos(6)

Key Documents

HPA021121

Sigma-Aldrich

Anti-MYO18A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-MAJN, Anti-Molecule associated with JAK3 N-terminus, Anti-Myosin containing a PDZ domain, Anti-Myosin-XVIIIa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

RFSFSQRSRDESASETSTPSEHSAAPSPQVEVRTLEGQLVQHPGPGIPRPGHRSRAPELVTKKFPVDLRLPPVVPLPPPTLRELELQRRPTGDFGFSLRRTTMLDRGPEGQACRRVVHFAEP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO18A(399687)

General description

The gene MYO18A (myosin-XVIIIa) is mapped to human chromosome 17q11.2. The protein contains two actin binding sites. It belongs to the myosin superfamily.

Immunogen

Myosin-XVIIIa recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MYO18A (myosin-XVIIIa) interacts with actin. MYO18A forms a tripartite complex with MRCK (myotonic dystrophy kinase-related CDC42-binding kinase) and LRAP35a (leucine repeat adapter protein 35A). The complex regulates lamellar actomyosin retrograde flow, cell protrusion and migration. MYO18A binds with GOLPH3 (golgi phosphoprotein 3) and thereby connects the Golgi to F-actin. MYO18A also interacts with PAK2 (p21-activated kinase 2)/PIXB (PAK-interacting exchange factor β)/GIT1 (GRK-interacting protein 1) complex, regulating epithelial cell migration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74261

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel H Taft et al.
The Journal of biological chemistry, 288(42), 30029-30041 (2013-08-31)
Molecular motors of the myosin superfamily share a generic motor domain region. They commonly bind actin in an ATP-sensitive manner, exhibit actin-activated ATPase activity, and generate force and movement in this interaction. Class-18 myosins form heavy chain dimers and contain
Young Ho Lee et al.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology, 36(1), 53-59 (2014-07-20)
The aims of this study were to identify candidate single nucleotide polymorphisms (SNPs) and mechanisms of Alzheimer's disease (AD) and to generate SNP to gene to pathway hypotheses. An AD genome-wide association study (GWAS) dataset that included 370,542 SNPs in
Rae-Mann Hsu et al.
Molecular biology of the cell, 21(2), 287-301 (2009-11-20)
The p21-activated kinase (PAK) 2 is known to be involved in numerous biological functions, including the regulation of actin reorganization and cell motility. To better understand the mechanisms underlying this regulation, we herein used a proteomic approach to identify PAK2-interacting
Paroma Chatterjee et al.
Journal of cell science, 136(10) (2023-04-25)
GIPC3 has been implicated in auditory function. Here, we establish that GIPC3 is initially localized to the cytoplasm of inner and outer hair cells of the cochlea and then is increasingly concentrated in cuticular plates and at cell junctions during
Ivan Tan et al.
Cell, 135(1), 123-136 (2008-10-16)
Actomyosin retrograde flow underlies the contraction essential for cell motility. Retrograde flow in both lamellipodia and lamella is required for membrane protrusion and for force generation by coupling to cell adhesion. We report that the Rac/Cdc42-binding kinase MRCK and myosin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service