Skip to Content
Merck
All Photos(3)

Key Documents

HPA021069

Sigma-Aldrich

Anti-TPP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TPP-II, Anti-Tripeptidyl aminopeptidase, Anti-Tripeptidyl-peptidase 2, Anti-Tripeptidyl-peptidase II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SSQANKLIKEELQSQVELLNSFEKKYSDPGPVYDCLVWHDGEVWRACIDSNEDGDLSKSTVLRNYKEAQEYGSFGTAEMLNYSVNIYDDGNLLSIVTSGGAHGTHVASIAAGHFPEEPERNGVAPGAQILSIKIGDTRLST

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPP2(7174)

General description

The gene TPP2 (tripeptidyl-peptidase 2) is mapped to human chromosome 13q32-q33. The protein mainly localizes in the cytoplasm, however it can translocate into the nucleus.

Immunogen

Tripeptidyl-peptidase 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-TPP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

TPP2 (tripeptidyl-peptidase 2) is a serine exopeptidase and participates in extralysosomal peptide degradation. Down-regulation of TPP2 is linked with early-onset Evans syndrome, immunodeficiency and premature immunosenescence. Major role of TPP2 is to maintain a steadiness between availability of intracellular amino acid, lysosome number and glycolysis. It is also suggested to have a role in DNA damage response. It is up-regulated in oral squamous cell carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74699

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulio Preta et al.
Biochemical and biophysical research communications, 389(4), 575-579 (2009-09-15)
Responses to DNA damage are influenced by cellular metabolism through the continuous production of reactive oxygen species (ROS), of which most are by-products of mitochondrial respiration. ROS have a strong influence on signaling pathways during responses to DNA damage, by
Polina Stepensky et al.
Blood, 125(5), 753-761 (2014-11-22)
Autoimmune cytopenia is a frequent manifestation of primary immunodeficiencies. Two siblings presented with Evans syndrome, viral infections, and progressive leukopenia. DNA available from one patient showed a homozygous frameshift mutation in tripeptidyl peptidase II (TPP2) abolishing protein expression. TPP2 is
Wei Lu et al.
Cell, 159(7), 1578-1590 (2014-12-20)
Proteasomes and lysosomes constitute the major cellular systems that catabolize proteins to recycle free amino acids for energy and new protein synthesis. Tripeptidyl peptidase II (TPPII) is a large cytosolic proteolytic complex that functions in tandem with the proteasome-ubiquitin protein degradation
T Martinsson et al.
Genomics, 17(2), 493-495 (1993-08-01)
We have assigned the human tripeptidyl peptidase II (TPP2) gene to chromosome region 13q32-q33 using two different methods. First, a full-length TPP2 cDNA was used as a probe on Southern blots of DNA from a panel of human/rodent somatic cell
Katsuya Usukura et al.
Journal of cancer research and clinical oncology, 139(1), 123-130 (2012-09-19)
Tripeptidyl peptidase II (TPP2), a member of the family of eukaryotic serine peptidase, has been implicated in DNA repair, cellular division, and apoptosis. The aim of this study was to examine TPP2 expression and its functional mechanisms in oral squamous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service