Skip to Content
Merck
All Photos(4)

Key Documents

HPA020959

Sigma-Aldrich

Anti-AGK antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acylglycerol kinase, mitochondrial, Anti-HsMuLK, Anti-MuLK, Anti-Multi-substrate lipid kinase, Anti-Multiple substrate lipid kinase, Anti-hAGK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
9
technique(s):
immunohistochemistry: 1:200- 1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LHLSGMDVTIVKTDYEGQAKKLLELMENTDVIIVAGGDGTLQEVVTGVLRRTDEATFSKIPIGFIPLGETSSLSHTLFAESGNKV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGK(55750)

Related Categories

General description

The gene AGK (acylglycerol kinase) is mapped to human chromsome 7q34. The protein localizes in the mitochondria. AGK is strongly expressed in the heart, muscle, kidney and brain.

Immunogen

Acylglycerol kinase, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-AGK antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AGK (acylglycerol kinase) is responsible for formation of lysophosphatidic acid (LPA) and phosphatidic acid (PA) through phosphorylation of monoacylglycerol and diacylglycerol, respectively. Additionally, AGK-mediated scavenging of mitochondrial diacylglycerol has been reported. It is up-regulated in prostate cancer and participates in amplification of EGF (epidermal growth factor) signaling. AGK also promotes tumorigenesis in breast cancer, esophageal squamous cell carcinoma and hepatocellular carcinoma. Mutations in AGK are linked with Sengers syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74837

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanmei Cui et al.
Oncotarget, 5(23), 12057-12069 (2014-12-05)
High levels of angiogenesis and resistance to apoptosis are major clinical features of hepatocellular carcinoma (HCC), a lethal disease with a high incidence worldwide. However, the precise mechanisms underlying these malignant properties remain unclear. Here, we demonstrated that acylglycerol kinase
Xi Wang et al.
Molecular cancer, 13, 106-106 (2014-06-03)
Acylglycerol kinase (AGK) is reported to be overexpressed in multiple cancers. The clinical significance and biological role of AGK in breast cancer, however, remain to be established. AGK expression in breast cancer cell lines, paired patient tissues were determined using
A Shiels et al.
Clinical genetics, 84(2), 120-127 (2013-05-08)
The pathogenesis of inherited cataracts of all kinds recapitulates the developmental and cell biology of the lens. Just as each novel mutation provides additional information about the structural or functional biology of the affected gene, each newly identified gene provides
Guijuan Liu et al.
Molecular medicine reports, 12(2), 2225-2230 (2015-04-16)
Cell proliferation is a major underlying cause of mortality amongst patients with oral squamous cell carcinoma (OSCC); however, the underlying mechanisms have remained to be elucidated. Acylglycerol kinase (AGK) is a multisubstrate lipid kinase, which is known to be associated
Ahmed M Abu El-Asrar et al.
Acta ophthalmologica, 90(2), e84-e89 (2011-11-23)
Lysophosphatidic acid (LPA)/LPA(1) receptor pathway is involved in inflammation, angiogenesis and fibrosis. This study was conducted to analyse the expression of LPA-producing enzymes, autotaxin (ATX) and acylglycerol kinase (AGK) and LPA(1) receptor, in proliferative diabetic retinopathy (PDR) and proliferative vitreoretinopathy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service