Skip to Content
Merck
All Photos(1)

Key Documents

HPA020107

Sigma-Aldrich

Anti-PLCD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1, Anti-PLC-III, Anti-PLC-delta-1, Anti-Phosphoinositide phospholipase C, Anti-Phospholipase C-delta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

EAFYKMLTQRVEIDRTFAEAAGSGETLSVDQLVTFLQHQQREEAAGPALALSLIERYEPSETAKAQRQMTKDGFLMYLLSADGSAFSLAHRRVYQDMGQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCD1(5333)

General description

Phospholipase C-δ1 (PLCD1) is an enzyme which is very sensitive to the changes in the levels of Ca2+. The gene encoding it is located on human chromosome 3p21.3-p22.

Immunogen

1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1 recombinant protein epitope signature tag (PrEST)

Application

Anti-PLCD1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Phospholipase C-δ1 (PLCD1) hydrolyzes phosphatidylinositol-4, 5-bisphosphate to form inositol 1, 4, 5 trisphosphate (IP3) and diacylglycerol, which are secondary messengers. It is involved in different pathways concerned with calcium homeostasis and metabolism. PLCD1 has an effect on the progression of the cell cycle in breast cancer cells and mutations in the gene encoding it has been linked to hereditary leukonychia. It has also been studied as a tumor suppressor gene for esophageal squamous cell carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74721

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hina Mir et al.
European journal of dermatology : EJD, 22(6), 736-739 (2012-11-15)
Hereditary leukonychia or porcelain nails is a nail dystrophy characterized by whitening of the nail plates in all nails of the hands and feet. It may exhibit an autosomal recessive or autosomal dominant pattern of inheritance. Mutations in the gene
Tingxiu Xiang et al.
Cancer biology & therapy, 10(5), 520-527 (2010-07-27)
Chromosome 3p harbors multiple tumor-suppressor genes. PLCD1, located at 3p22, encodes an enzyme that mediates regulatory signaling of energy metabolism, calcium homeostasis and intracellular movement. We investigated the epigenetic alterations of PLCD1 and its tumor suppressor function in breast cancer.
Haixi Mu et al.
Experimental cell research, 332(2), 179-189 (2015-02-07)
Our previous study showed that PLCD1 significantly decreases cell proliferation and affects cell cycle progression in breast cancer cells. In the present study, we aimed to investigate its functional and molecular mechanisms, and whether or not can become a new
X-T Hu et al.
Oncogene, 28(26), 2466-2475 (2009-05-19)
Located at the important tumor suppressor locus, 3p22, PLCD1 encodes an enzyme that mediates regulatory signaling of energy metabolism, calcium homeostasis and intracellular movements. We identified PLCD1 as a downregulated gene in aerodigestive carcinomas through expression profiling and epigenetic characterization.
Shuji Shibutani et al.
Circulation, 125(8), 1027-1036 (2012-01-24)
We reported that phospholipase C (PLC)-δ1 activity was enhanced 3-fold in patients with coronary spastic angina. We detected variant PLC-δ1 with replacement of arginine 257 by histidine (R257H) showing increased enzymatic activity. We tested the hypothesis that increased PLC-δ1 activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service