Skip to Content
Merck
All Photos(5)

Key Documents

HPA019879

Sigma-Aldrich

Anti-URGCP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-AC004985.2-1, Anti-HBV X protein up-regulated gene 4 protein, Anti-HBxAg up-regulated gene 4 protein, Anti-Protein URG4, Anti-URG4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PSLSEKQYFLRWMEWGLARVAQPRLRQPPETLLTLRPKHGGTTDVGEPLWPEPLGVEHFLREMGQFYEAESCLVEAGRLPAGQRRFAHFP

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... URGCP(55665)

General description

Upregulator of cell proliferation (URGCP) is a 104kDa, plasma membrane localized protein. It is mapped on chromosome 7p13.

Immunogen

Protein URG4 recombinant protein epitope signature tag (PrEST)

Application

Anti-URGCP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Upregulator of cell proliferation (URGCP) is associated with the onset of oncogenesis and cell cycle regulation. Its overexpression has been observed in the hepatitis B-infected liver, gastric cancer cells. It also alters cyclin D1 expression in cell proliferation stage. It has been reported that URGCP may be used as a therapeutic target for the treatment of cervical cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74938

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiping Li et al.
Archives of gynecology and obstetrics, 286(1), 209-215 (2012-03-08)
Up-regulated gene 4 (URG4) has been demonstrated to be involved in progression of various human cancers. This study investigated the clinicopathological significance of URG4 in epithelial ovarian cancer (EOC). Immunohistochemistry was applied to investigate the expression of URG4 in ovarian
N Lale Satiroglu Tufan et al.
Neoplasia (New York, N.Y.), 4(4), 355-368 (2002-06-26)
Hepatitis B virus encoded X antigen (HBxAg) may contribute to the development of hepatocellular carcinoma (HCC) by up- or downregulating the expression of cellular genes that promote cell growth and survival. To test this hypothesis, HBxAg-positive and -negative HepG2 cells
Lan Zhang et al.
BMC cancer, 14, 885-885 (2014-11-28)
Upregulator of cell proliferation 4 (URG4) has been implicated in the oncogenesis of certain cancers. However, the correlation between URG4 expression and clinicopathological significance in human cancer remains unclear. Therefore, this study investigated its expression and clinicopathological significance in cervical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service