Skip to Content
Merck
All Photos(10)

Key Documents

HPA019828

Sigma-Aldrich

Anti-AMPH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Amphiphysin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

EKVIPSVVIEPASNHEEEGENEITIGAEPKETTEDAAPPGPTSETPELATEQKPIQDPQPTPSAPAMGAADQLASAREA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMPH(273)

General description

AMPH (Amphiphysin) is a 128kD peripheral protein highly distributed in nervous system including many neurons, certain endocrine cell types, and spermatocytes. It exists in two isoforms with two different molecular weight 128 and 108kD in the breast cancer. In human, it is located on chromosome 7, region p13-p14. It is a member of amphiphysin family and consists of SH3 domain.

Immunogen

Amphiphysin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AMPH (Amphiphysin) is involved in several pleotropic functions including roles in endocytosis, actin function, and cell growth control. It plays an important role in synaptic vesicle endocytosis of axon terminals. Alteration in AMPH gene is associated with heritable disorders of the nervous system, endocrine tissues or male fertility. It also has correlation with a rare neurologic autoimmune disease, paraneoplastic Stiff-Man syndrome. It is also invloved in neoplastic growth cone dynamics.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74867

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Yamamoto et al.
Human molecular genetics, 4(2), 265-268 (1995-02-01)
Amphiphysin is a protein peripherally associated with synaptic vesicles. It is expressed in many neurons, certain endocrine cell types, and spermatocytes. Autoantibodies against amphiphysin occur in patients afflicted with a rare neurologic autoimmune disease, paraneoplastic Stiff-Man syndrome. To provide a
S Floyd et al.
Molecular medicine (Cambridge, Mass.), 4(1), 29-39 (1998-03-26)
Amphiphysin I is a 128 kD protein highly concentrated in nerve terminals, where it has a putative role in endocytosis. It is a dominant autoantigen in patients with stiff-man syndrome associated with breast cancer, as well as in other paraneoplastic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service