Skip to Content
Merck
All Photos(6)

Key Documents

HPA019652

Sigma-Aldrich

Anti-POU2F3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Octamer-binding transcription factor 11, Anti-PLA-1 protein, Anti-POU domain, class 2, transcription factor 3, Anti-Transcription factor Skn-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQFLLSQTQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POU2F3(25833)

General description

POU class 2 homeobox 3 (POU2F3) is located on human chromosome location 11q23.3.
POU2F3 (POU class 2 homeobox 3) is a homeodomain protein, important for the generation of sweet, umami and bitter taste cells. Specifically, it is localized in stratified squamous epithelia, including the epidermis, cervix and foreskin.

Immunogen

POU domain, class 2, transcription factor 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-POU2F3 antibody produced in rabbit has been used in:
  • western blotting
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

POU2F3 (POU class 2 homeobox 3) is a keratinocyte-specific POU transcription factor, which is majorly responsible for the generation and maintenance of solitary chemosensory cells such as sweet, umami and bitter taste cells. POU2F3 is a tumor suppressor candidate gene in cervical cancer (CC) since; its expression has been found in heterozygosity region on11q23.3. It also participates in keratinocyte proliferation and terminal differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86702

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypermethylation of the CpG island of POU domain, class 2, transcription factor 3 (POU2F3) in human cervical cancer and cell lines
Zhang Z, et al.
Cancer Research, 65, 217-217 (2005)
POU2F3 is a master regulator of a tuft cell-like variant of small cell lung cancer
Huang YH, et al.
Genes & Development, 32(13-14), 915-928 (2018)
Intestinal epithelial tuft cells initiate type 2 mucosal immunity to helminth parasites
Gerbe F, et al.
Nature, 529(7585), 226-226 (2016)
Jin Ng et al.
Methods in molecular biology (Clifton, N.J.), 2806, 117-138 (2024-04-28)
Unlocking the heterogeneity of cancers is crucial for developing therapeutic approaches that effectively eradicate disease. As our understanding of markers specific to cancer subclones or subtypes expands, there is a growing demand for advanced technologies that enable the simultaneous investigation
François Gerbe et al.
Nature, 529(7585), 226-230 (2016-01-15)
Helminth parasitic infections are a major global health and social burden. The host defence against helminths such as Nippostrongylus brasiliensis is orchestrated by type 2 cell-mediated immunity. Induction of type 2 cytokines, including interleukins (IL) IL-4 and IL-13, induce goblet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service