Skip to Content
Merck
All Photos(3)

Key Documents

HPA019502

Sigma-Aldrich

Anti-S100P antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein S100-P, Anti-S100 calcium-binding protein P

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:2500- 1:5000

immunogen sequence

MTELETAMGMIIDVFSRYSGSEGSTQTLTKGELKVLMEKELPGFLQSGKDKDAVDKLLKDLDANGDAQVDFSEF

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100P(6286)

General description

S100P (S100 calcium binding protein P) is a calcium binding protein belonging to the S100 gene family. It is expressed in the human placenta. Apart from normal cells, its expression has been found in several malignant neoplasms.

Immunogen

Protein S100-P recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

S100P (S100 calcium binding protein P) is associated with several processes such as calcium homeostasis, cell proliferation, apoptosis, cell invasion and motility. It functions as a Ca2+ signal transducer to activate the membrane as well as F-actin binding protein ezrin during Ca2+ bound conformation. It can also facilitate cell proliferation and survival via receptor for activated glycation end products (RAGE). It has been reported that S100P can be a potential prognostic marker for the triple-negative breast cancer (TNBC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74602

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judith Austermann et al.
Biochimica et biophysica acta, 1793(6), 1078-1085 (2008-12-30)
S100 proteins function as Ca2+ signal transducers by regulating cellular targets in their Ca2+ bound conformation. S100P is a member of the S100 protein family that can activate the membrane and F-actin binding protein ezrin in a Ca2+ dependent manner
Melanie Maierthaler et al.
Experimental and molecular pathology, 99(1), 180-187 (2015-06-27)
Triple-negative breast cancer (TNBC) is a group of very aggressive breast tumours, characterised by lack of expression of oestrogen receptor (ER), progesterone receptor (PR) and erb-b2 receptor tyrosine kinase 2 (ERBB2/HER2). Nevertheless, TNBCs show different clinical characteristics and are very
Thiruvengadam Arumugam et al.
The Journal of biological chemistry, 279(7), 5059-5065 (2003-11-18)
S100P is a member of the S100 protein family that is expressed in several malignant neoplasms. Currently the effects of this molecule on cell function are unknown. In the present study we investigated the biological effects and mechanisms of action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service