Skip to Content
Merck
All Photos(9)

Key Documents

HPA019484

Sigma-Aldrich

Anti-BAIAP2L1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-BAI1-associated protein 2-like protein 1, Anti-Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1, Anti-Insulin receptor tyrosine kinase substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

ERSNVVRKDYDTLSKCSPKMPPAPSGRAYTSPLIDMFNNPATAAPNSQRVNNSTGTSEDPSLQRSVSVATGLNMMKKQKVKTIFPHTAGSN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BAIAP2L1 (BAI1-associated protein 2-like 1) is a 53kDa scaffold protein of a newly discovered family. It is involved in the signal transducing and control of cytoskeleton assembly as well as membrane protrusion at the plasma membrane. Tyrosine phosphorylated BAIAP2L1 helps to deform the plasma membrane structure, which is an essential step in cell motility. BAIAP2L1 also has clinical association in the pathogenesis of malignant tumors, including hepatocellular carcinoma (HCC). It has been studied that during proliferation of HCC, BAIAP2L1 interacts with epidermal growth factor receptor (EGFR) to phosphorylate extracellular signal-regulated kinase (ERK) via EGFR-ERK signalling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74833

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Ping Wang et al.
Cancer letters, 337(1), 96-106 (2013-05-23)
Insulin receptor tyrosine kinase substrate (IRTKS) is closely associated with actin remodelling and membrane protrusion, but its role in the pathogenesis of malignant tumours, including hepatocellular carcinoma (HCC), is still unknown. In this study, we showed that IRTKS was frequently
Giorgio Scita et al.
Trends in cell biology, 18(2), 52-60 (2008-01-25)
A tight spatiotemporal coordination of the machineries controlling membrane bending and trafficking, and actin dynamics is crucial for the generation of cellular protrusions. Proteins that are simultaneously capable of regulating actin dynamics and sensing or inducing membrane curvature are predicted
J Michael Henderson et al.
The EMBO journal, 42(24), e113761-e113761 (2023-11-27)
Tunnelling nanotubes (TNTs) connect distant cells and mediate cargo transfer for intercellular communication in physiological and pathological contexts. How cells generate these actin-mediated protrusions to span lengths beyond those attainable by canonical filopodia remains unknown. Through a combination of micropatterning
Ai Mei Chou et al.
Scientific reports, 7, 40485-40485 (2017-01-10)
The insulin receptor substrate of 53 kDa, IRSp53, is an adaptor protein that works with activated GTPases, Cdc42 and Rac, to modulate actin dynamics and generate membrane protrusions in response to cell signaling. Adult mice that lack IRSp53 fail to regulate
Gang Chen et al.
FEBS letters, 585(19), 2972-2978 (2011-08-16)
Insulin receptor tyrosine kinase substrate (IRTKS) has been demonstrated to be a scaffold protein involved in plasma membrane deformation and actin cytoskeleton remodeling. IRTKS is tyrosine phosphorylated in response to insulin stimulation. However, the mechanism and function of IRTKS phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service