Skip to Content
Merck
All Photos(3)

Key Documents

HPA019195

Sigma-Aldrich

Anti-HCN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BCNG-1, Anti-BCNG1, Anti-HAC-2, Anti-hyperpolarization activated cyclic nucleotide-gated potassium channel 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
6
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PSLPHEVSTLISRPHPTVGESLASIPQPVTAVPGTGLQAGGRSTVPQRVTLFRQMSSGAIPPNRGVPPAPPPPAAALPRESSSVLNTDPDAE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HCN1(348980)

General description

The gene HCN1 (potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1) is mapped to human chromosome 5p12. The protein contains transmembrane domains and a carboxy-terminal cyclic nucleotide-binding domain. HCN1 is strongly expressed in the plasma membrane of neurons and is also present in particular compartments (inner segments of photoreceptors, stereocilia of cochlear hair cells).

Immunogen

hyperpolarization activated cyclic nucleotide-gated potassium channel 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HCN (potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel) channels get activated by hyperpolarization and can be modulated by cyclic-AMP. They cause cationic ′Ih′ current in neurons, resulting in neuronal excitability and signal transmission in the nervous system. HCN channels mainly carry inward currents of K+ and Na+. Mutations in HCN1 are linked with early infantile epileptic encephalopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72946

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Funny current and sudden cardiac death
Perde F
Romanian Journal of legal medicine, 23, 95-100 (2015)
Caroline Nava et al.
Nature genetics, 46(6), 640-645 (2014-04-22)
Hyperpolarization-activated, cyclic nucleotide-gated (HCN) channels contribute to cationic Ih current in neurons and regulate the excitability of neuronal networks. Studies in rat models have shown that the Hcn1 gene has a key role in epilepsy, but clinical evidence implicating HCN1
B Santoro et al.
Cell, 93(5), 717-729 (1998-06-18)
The generation of pacemaker activity in heart and brain is mediated by hyperpolarization-activated cation channels that are directly regulated by cyclic nucleotides. We previously cloned a novel member of the voltage-gated K channel family from mouse brain (mBCNG-1) that contained
Catherine H Gill et al.
British journal of pharmacology, 143(3), 411-421 (2004-09-08)
The human hyperpolarization-activated cyclic nucleotide-gated 1 (hHCN1) subunit was heterologously expressed in mammalian cell lines (CV-1 and CHO) and its properties investigated using whole-cell patch-clamp recordings. Activation of this recombinant channel, by membrane hyperpolarization, generated a slowly activating, noninactivating inward
Ye Han et al.
The Journal of biological chemistry, 286(23), 20823-20834 (2011-04-21)
Ion channel trafficking and gating are often influenced by interactions with auxiliary subunits. Tetratricopeptide repeat-containing Rab8b-interacting protein (TRIP8b) is an auxiliary subunit for neuronal hyperpolarization-activated cyclic nucleotide-gated (HCN) channels. TRIP8b interacts directly with two distinct sites of HCN channel pore-forming

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service