Skip to Content
Merck
All Photos(7)

Key Documents

HPA019092

Sigma-Aldrich

Anti-CAPG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3

Synonym(s):

Anti-Actin regulatory protein CAP-G, Anti-Macrophage-capping protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

ERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVAD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPG(822)

General description

The gene CAPG (macrophage-capping gene) is mapped to human chromosome 2p11.2. It belongs to gelsolin protein family. The protein localizes in the cytoplasm and nucleus.

Immunogen

Macrophage-capping protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CAPG (macrophage-capping protein) is involved in regulation of actin length via capping the plus or barbed ends in a Ca2+- and PIP2 (phosphatidylinositol 4,5-bisphosphate)-dependent manner. It is also identified as an oncogene. CAPG is up-regulated in breast cancer, ovarian cancer, hepatocellular carcinoma and lung cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74673

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Glaser et al.
BioMed research international, 2014, 379847-379847 (2014-05-08)
The actin binding protein CapG modulates cell motility by interacting with the cytoskeleton. CapG is associated with tumor progression in different nongynecologic tumor entities and overexpression in breast cancer cell lines correlates with a more invasive phenotype in vitro. Here
Kyung Ran Jun et al.
Molecular cytogenetics, 7, 52-52 (2014-10-09)
Copy number variations at 2p11.2 have been rare and to our knowledge, no abnormal phenotype with an interstitial 2p11.2 duplication has yet been reported. Here we report the first case with syndromic intellectual disability associated with microduplication at 2p11.2. We
Fangchun Shao et al.
The Tohoku journal of experimental medicine, 225(2), 95-101 (2011-09-13)
Gelsolin-like actin-capping protein (CapG), a ubiquitous actin-binding protein, has been shown to play a critical role in regulating the migration ability of cells. In this study, we investigated CapG expression in lung cancer cell lines under hypoxia and evaluated the
Kazuya Kimura et al.
Journal of proteomics, 78, 362-373 (2012-10-23)
Hepatocellular carcinoma (HCC) is one of the most deadly cancers worldwide. We performed a proteomic study to understand the molecular mechanisms underlying metastasis in HCC. Among the 3491 protein spots observed by two-dimensional difference gel electrophoresis (2D-DIGE), we found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service