Skip to Content
Merck
All Photos(4)

Key Documents

HPA018976

Sigma-Aldrich

Anti-PRMT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein arginine N-methyltransferase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LRFDIRKAGTLHGFTAWFSVHFQSLQEGQPPQVLSTGPFHPTTHWKQTLFMMDDPVPVHTGDVVTGSVVLQRNPVWRRHMSVALSW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRMT2(3275)

General description

The gene PRMT2 (protein arginine N-methyltransferase 2) is mapped to human chromosome 21q22.3. It belongs to the arginine methyltransferase protein family. The protein shuttles between the cytoplasm and nucleus. PRMT2 is also referred as HRMT1L1 (histone-arginine N-methyltransferase).

Immunogen

Protein arginine N-methyltransferase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRMT2 (protein arginine N-methyltransferase 2) methylates arginine residues within proteins using S-adenosyl-L-methionine. It methylates histone H4 and acts as a transcription co-activator. PRMT2 is a co-activator of the androgen receptor and ERα (Estrogen erceptor α). PRMT2 interacts with PRMT1 and promotes PRMT1 enzymatic activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73894

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Poelmans et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 150B(1), 140-147 (2008-06-04)
Dyslexia is the most common childhood learning disorder and it is a significantly heritable trait. At least nine chromosomal loci have been linked to dyslexia, and additional susceptibility loci on other chromosomes have been suggested. Within two of these loci
Rene Meyer et al.
The Journal of steroid biochemistry and molecular biology, 107(1-2), 1-14 (2007-06-26)
The basal transcriptional activity of nuclear receptors (NRs) is regulated by interactions with additional comodulator proteins (coactivator/corepressor). Here, we describe a new androgen receptor (AR)-associated coactivator, PRMT2, which belongs to the arginine methyltransferase protein family. To search for AR-interacting proteins
Chao Qi et al.
The Journal of biological chemistry, 277(32), 28624-28630 (2002-06-01)
In an attempt to isolate cofactors capable of influencing estrogen receptor alpha (ERalpha) transcriptional activity, we used yeast two-hybrid screening and identified protein arginine methyltransferase 2 (PRMT2) as a new ERalpha-binding protein. PRMT2 interacted directly with three ERalpha regions including
Magnolia L Pak et al.
Biochemistry, 50(38), 8226-8240 (2011-08-20)
Protein arginine N-methyltransferases (PRMTs) act in signaling pathways and gene expression by methylating arginine residues within target proteins. PRMT1 is responsible for most cellular arginine methylation activity and can work independently or in collaboration with other PRMTs. In this study
Ted M Lakowski et al.
The Biochemical journal, 421(2), 253-261 (2009-05-02)
Protein arginine N-methyltransferases (PRMTs) methylate arginine residues within proteins using S-adenosyl-L-methionine (AdoMet) to form S-adenosyl-L-homocysteine and methylarginine residues. All PRMTs produce omega-NG-monomethylarginine (MMA) residues and either asymmetric omega-N(G),N(G)-dimethylarginine (aDMA) or symmetric omega-N(G),N'(G)-dimethylarginine (sDMA) residues, referred to as Type I or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service