Skip to Content
Merck
All Photos(1)

Key Documents

HPA018951

Sigma-Aldrich

Anti-BAG4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BAG family molecular chaperone regulator 4, Anti-BAG-4, Anti-Bcl-2-associated athanogene 4, Anti-Silencer of death domains

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

VHQYESSGTVNNDDSDLLDSQVQYSAEPQLYGNATSDHPNNQDQSSSLPEECVPSDESTPPSIKKIIHVLEKVQYLEQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAG4(9530)

General description

The gene BAG4 (Bcl associated anthanogene family molecular chaperone regulator 4) is mapped to human chromosome 8q12. It is widely expressed. BAG4 localizes in the cytoplasm and the nucleus. BAG4 is also referred as SODD (silencer of death domains).

Immunogen

BAG family molecular chaperone regulator 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BAG4 (Bcl associated anthanogene family molecular chaperone regulator 4) functions as an adaptor and interacts with Hsp (Heat shock protein)-90/Hsc70. It interacts with tumor necrosis factor receptor type-1 (TNF-R1) and negatively regulates the apoptosis-inducing signaling pathway mediated by TNF-R1. It is also a regulator of CD95 activity. In addition, BAG4 binds to apobec-1 (apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1) and negatively regulates apobec-1-mediated apolipoprotein B mRNA editing. BAG4 also protects melanoma cells from apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74213

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hedda Eichholtz-Wirth et al.
Cancer letters, 194(1), 81-89 (2003-04-23)
The involvement of SODD/BAG-4 was studied in TNFR1 signaling, using SODD/BAG-4- overexpressing HeLa cells as a cellular model. Stable transfection of a SODD/BAG-4 cDNA leads to increased levels of the full-length 70-kDa protein and additional C-terminal fragments as well as
Y Jiang et al.
Science (New York, N.Y.), 283(5401), 543-546 (1999-01-23)
Tumor necrosis factor receptor type 1 (TNF-R1) contains a cytoplasmic death domain that is required for the signaling of TNF activities such as apoptosis and nuclear factor kappa B (NF-kappaB) activation. Normally, these signals are generated only after TNF-induced receptor
H F Tao et al.
Genetics and molecular research : GMR, 13(1), 2020-2031 (2014-04-17)
This study explored the clinical significance of silencer of death domain (SODD) expression in childhood acute lymphoblastic leukemia (ALL) and its influence on chemotherapy as well as the effect of SODD expression on apoptosis of leukemic cells. The expression of
Paul P Lau et al.
The Journal of biological chemistry, 278(52), 52988-52996 (2003-10-16)
Apobec-1 is the catalytic subunit of a multicomponent editosome complex that mediates apolipoprotein B (apoB) mRNA editing. We isolated a novel apobec-1-interacting protein by yeast two-hybrid cloning and identified the protein as BAG-4. BAG-4, a chaperone-regulating protein, also known as
Klara Briknarová et al.
The Journal of biological chemistry, 277(34), 31172-31178 (2002-06-12)
BAG (Bcl-2-associated athanogene) proteins are molecular chaperone regulators that affect diverse cellular pathways. All members share a conserved motif, called the BAG domain (BD), which binds to Hsp70/Hsc70 family proteins and modulates their activity. We have determined the solution structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service