Skip to Content
Merck
All Photos(1)

Key Documents

HPA018893

Sigma-Aldrich

Anti-NSD3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ20353, Anti-KMT3F, Anti-WHISTLE, Anti-WHSC1L1, Anti-Histone-lysine N-methyltransferase NSD3, Anti-Nuclear SET domain-containing protein 3, Anti-WHSC1-like protein 1, Anti-Whistle, Anti-Wolf-Hirschhorn syndrome candidate 1-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

NTQPEQTNAGEVASSLSSTEIRRHSQRRHTSAEEEEPPPVKIAWKTAAARKSLPASITMHKGSLDLQKCNMSPVVKIEQVFALQNATGDGKFIDQFVYSTKGIGNKTEISVRGQDRLIISTPNQRNEKPTQSVSSPEATS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WHSC1L1(54904)

General description

The gene WHSC1L1 (Wolf-Hirschhorn syndrome candidate 1-like protein 1) is mapped to human chromosome 8p11.2. The protein contains methyltransferase SET domain and the PHD (plant homeodomain) finger. WHSC1L1 is also called as NSD3 (nuclear SET domain-containing protein 3).

Immunogen

nuclear receptor binding SET domain protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Wolf-Hirschhorn syndrome candidate 1-like protein 1 (WHSC1L1) is a histone methyltransferase. Bromodomain protein 4 (BRD4) interacts with WHSC1L1 and thereby regulates transcriptional activity. WHSC1L1 modulates transcription by controlling histone-3 lysine-36 (H3K36) methylation. WHSC1L1 is up-regulated in tumor cell lines, bladder, lung, liver cancer and primary breast carcinoma. It is important for cell cycle progression at mitosis. WHSC1L1-NUT (nuclear protein in testis) fusion protein is associated with NUT (nuclear protein of the testis) midline carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85172

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P O Angrand et al.
Genomics, 74(1), 79-88 (2001-05-26)
We describe the isolation and characterization of NSD3, the third member of a gene family including Nsd1 and NSD2. Murine Nsd1 was isolated in a search for proteins that interact with the ligand-binding domain of retinoic acid receptor alpha. NSD2
Daechun Kang et al.
Genes, chromosomes & cancer, 52(2), 126-139 (2012-09-27)
Histone lysine methylation plays a fundamental role in chromatin organization. Although a set of histone methyltransferases have been identified and biochemically characterized, the pathological roles of their dysfunction in human cancers are still not well understood. In this study, we
Shaila Rahman et al.
Molecular and cellular biology, 31(13), 2641-2652 (2011-05-11)
Bromodomain protein 4 (Brd4) plays critical roles in development, cancer progression, and virus-host pathogenesis. To gain mechanistic insight into the various biological functions of Brd4, we performed a proteomic analysis to identify and characterize Brd4-associated cellular proteins. We found that
Christopher A French et al.
Cancer discovery, 4(8), 928-941 (2014-05-31)
NUT midline carcinoma (NMC) is an aggressive subtype of squamous cell carcinoma that typically harbors BRD4/3-NUT fusion oncoproteins that block differentiation and maintain tumor growth. In 20% of cases, NUT is fused to uncharacterized non-BRD gene(s). We established a new
I Stec et al.
Genomics, 76(1-3), 5-8 (2001-09-11)
We have identified and characterized a gene (60% on protein level) and a pseudogene (93% on DNA level) that show high similarity to the Wolf-Hirschhorn syndrome candidate gene-1 (WHSC1). These genes, WHSC1L1 and WHSC1L2P, map to human chromosomes 8p11.2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service