Skip to Content
Merck
All Photos(3)

Key Documents

HPA018852

Sigma-Aldrich

Anti-C1S antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C1 esterase, Anti-Complement C1s subcomponent precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AAHVVEGNREPTMYVGSTSVQTSRLAKSKMLTPEHVFIHPGWKLLEVPEGRTNFDNDIALVRLKDPVKMGPTVSPICLPGTSSDYNLMDGDLGLISGWGRTEKRDRAVRLKAARLPVAPLRKCKEVKVEKPTADAEAYVFTPNMICAGGE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1S(716)

General description

The gene complement component 1 subcomponent s (C1S) is mapped to human chromosome 12p13.2. The protein contains a CUB1 (C1r/C1s, Uegf, Bmp1)-EGF (Epidermal growth factor-like)-CUB2 region, two complement control protein modules (CCP1 and CCP2) and a serine protease (SP) domain.

Immunogen

Complement C1s subcomponent precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-C1S antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. The antibody has been used for bead array immunoassay.

Biochem/physiol Actions

Complement component 1 subcomponent s (C1S) is a subcomponent of C1, the first component of the complement system. The complement system is a first line of innate immune defense in cells. C1S is a serine protease. Activated C1S participates in immune response by binding and cleaving the effector molecule complement component-4 (C4). The CUB1 (C1r/C1s, Uegf, Bmp1)-EGF (Epidermal growth factor-like) region of C1S is responsible for mediating interaction between C1S and other components of C1 (C1R and C1Q). Absence of C1S is associated with autoimmune diseases, for instance lupus-like syndrome, Hashimoto′s thyroiditis, and autoimmune hepatitis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78220

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Perry et al.
The Journal of biological chemistry, 288(22), 15821-15829 (2013-04-18)
The complement system is an ancient innate immune defense pathway that plays a front line role in eliminating microbial pathogens. Recognition of foreign targets by antibodies drives sequential activation of two serine proteases, C1r and C1s, which reside within the
P R Lawson et al.
International immunology, 12(3), 231-240 (2000-03-04)
The complement system is a cascade of serum proteins and receptors which forms a vital arm of innate immunity and enhances the adaptive immune response. This work establishes the chromosomal localization of four key genes of the murine complement system.
Liisa Nissinen et al.
Scientific reports, 14(1), 13465-13465 (2024-06-13)
Cutaneous squamous cell carcinoma (cSCC) is the most common metastatic skin cancer. The incidence of cSCC is increasing globally and the prognosis of metastatic disease is poor. Currently there are no specific targeted therapies for advanced or metastatic cSCC. We
M A Dragon-Durey et al.
Journal of immunology (Baltimore, Md. : 1950), 166(12), 7612-7616 (2001-06-08)
We have investigated the molecular basis of selective and complete C1s deficiency in 2-year-old girl with complex autoimmune diseases including lupus-like syndrome, Hashimoto's thyroiditis, and autoimmune hepatitis. This patient's complement profile was characterized by the absence of CH50 activity, C1
Anamika Gangwar et al.
Scientific reports, 10(1), 7899-7899 (2020-05-15)
Intermittent hypoxia, initially associated with adverse effects of sleep apnea, has now metamorphosed into a module for improved sports performance. The regimen followed for improved sports performance is milder intermittent hypoxic training (IHT) as compared to chronic and severe intermittent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service