Skip to Content
Merck
All Photos(6)

Key Documents

HPA018844

Sigma-Aldrich

Anti-LIF antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-D factor, Anti-Differentiation- stimulating factor, Anti-Emfilermin, Anti-LIF, Anti-Leukemia inhibitory factor precursor, Anti-MLPLI, Anti-Melanoma-derived LPL inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MKVLAAGVVPLLLVLHWKHGAGSPLPITPVNATCAIRHPCHNNLMNQIRSQLAQLNGSANALFILYYTAQGEPFPNNLDKLCGPNVTDFPPFHANGTEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIF(3976)

General description

The gene leukemia inhibitory factor (LIF) is mapped to human chromosome 22q12. It belongs to IL (Interleukin)-6 proinflammatory cytokine family. LIF transcripts are present in all tissues except the brain, testis, skeletal muscle and peripheral blood leukocytes.

Immunogen

Leukemia inhibitory factor precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Leukemia inhibitory factor (LIF) is a cytokine. In fibroblasts, LIF causes carcinoma cell invasion through induction of transforming growth factor-β-dependent actomyosin contractility and extracellular matrix remodeling. Presence of LIF guards lungs from pathology during respiratory syncytial virus infection. LIF is also involved in stimulating osteoblast differentiation and reducing adipocyte differentiation by working in co-operation with protein kinase inhibitor-γ.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74569

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chrysoula Margioula-Siarkou et al.
Molecular medicine reports, 13(6), 4721-4728 (2016-04-16)
The aim of the present study was to determine the expression of leukemia inhibitory factor (LIF) and LIF receptor (LIF‑R) in the endometrium of fertile and infertile women during the implantation window. A prospective study was conducted between March 2013 and March 2015
E B Cullinan et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(7), 3115-3120 (1996-04-02)
The uterine expression of leukemia inhibitory factor (LIF) is essential for embryo implantation in the mouse. Here, we describe the expression of LIF, related members of this group of cytokines, oncostatin M and ciliary neurotrophic factor, and the LIF receptor
Robert F Foronjy et al.
BMC immunology, 15, 41-41 (2014-10-04)
Respiratory syncytial virus (RSV) infects the lung epithelium where it stimulates the production of numerous host cytokines that are associated with disease burden and acute lung injury. Characterizing the host cytokine response to RSV infection, the regulation of host cytokines
Jean Albrengues et al.
Cell reports, 7(5), 1664-1678 (2014-05-27)
Signaling crosstalk between tumor cells and fibroblasts confers proinvasive properties to the tumor microenvironment. Here, we identify leukemia inhibitory factor (LIF) as a tumor promoter that mediates proinvasive activation of stromal fibroblasts independent of alpha-smooth muscle actin (α-SMA) expression. We
G R Sutherland et al.
Leukemia, 3(1), 9-13 (1989-01-01)
The gene for human leukemia inhibitory factor (LIF) has been mapped by Southern analysis of a series of mouse/human somatic cell hybrids and by in situ hybridization to the chromosomes of two normal males and some individuals with chromosomal rearrangements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service