Skip to Content
Merck
All Photos(3)

Key Documents

HPA018265

Sigma-Aldrich

Anti-SYF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CBPIN, Anti-DKFZp564O2082, Anti-NTC31, Anti-SYF2 homolog, RNA splicing factor (S. cerevisiae), Anti-fSAP29, Anti-p29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

AAIAASEVLVDSAEEGSLAAAAELAAQKREQRLRKFRELHLMRNEARKLNHQEVVEEDKRLKLPANWEAKKARLEWELKEEEKKKECAARG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYF2(25949)

General description

Pre-mRNA-splicing factor SYF2 is abundantly expressed in heart, skeletal muscle and kidney. The protein is mainly localized in the nucleus.

Immunogen

SYF2 homolog, RNA splicing factor (S. cerevisiae) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pre-mRNA-splicing factor SYF2 is considered as a cell cycle regulator at the G1-S transition. The protein interacts with cyclin D-type binding-protein-1. SYF2 also associates with chromatin and interacts with DNA replication licensing factor MCM3 (mini-chromosome maintenance 3). Depletion of SYF2 down-regulates expression of PCNA (Proliferating cell nuclear antigen), cyclin D, inhibits DNA replication and arrest cell growth at G1 phase. SYF2 is up-regulated in human glioma cells and esophageal squamous cell carcinoma tumor tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72594

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junya Zhu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10275-10285 (2014-07-19)
SYF2, also known as CCNDBP1-interactor or p29, is reported in pre-mRNA splicing and cell cycle progression. However, the role of SYF2 in esophageal squamous cell carcinoma (ESCC) development remains elusive. In the present study, Western blot and immunohistochemistry assays demonstrated
Jun Guo et al.
Medical oncology (Northwood, London, England), 31(8), 101-101 (2014-07-06)
SYF2 is thought to be a cell cycle regulator at the G1/S transition, which encodes a nuclear protein that interacts with cyclin D-type binding-protein 1. In the present study, we investigated the role of SYF2 in human glioma progression. Immunohistochemical
Po-Chen Chu et al.
Cancer research, 66(17), 8484-8491 (2006-09-05)
Human p29 is a newly identified nuclear protein whose function is largely undetermined. We found that p29 associated with chromatin, interacted with MCM3, and localized with proliferating cell nuclear antigen foci in the S phase. Silencing of p29 using small
M S Chang et al.
Biochemical and biophysical research communications, 279(2), 732-737 (2000-12-19)
GCIP, a newly identified cyclin D-interacting protein, was found to reduce the phosphorylation of retinoblastoma protein and inhibit E2F1-mediated transcriptional activity. To explore more GCIP interacting proteins, the yeast two-hybrid screening using GCIP as a bait protein was performed. One

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service