Skip to Content
Merck
All Photos(3)

Key Documents

HPA017653

Sigma-Aldrich

Anti-DNAJC14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DRiP78, Anti-DnaJ homolog subfamily C member 14, Anti-DnaJ protein homolog 3, Anti-Dopamine receptor-interacting protein of 78 kDa, Anti-HDJ-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

WGWLELPWVKQNINRQGNAPVASGRYCQPEEEVARLLTMAGVPEDELNPFHVLGVEATASDVELKKAYRQLAVMVHPDKNHHPRAEEAFKVLRAAWDIVSNAEKRKEYEMKRMAENELSRSVNEFLSKLQDDLKEAMNTMMCSRCQG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAJC14(85406)

General description

DNAJC14 (DnaJ heat shock protein family (Hsp40) member C14) belongs to Hsp40 chaperone family. The protein contains 70-amino acid motif called J-domain and a C- terminal domain. J-domain facilitates ATP hydrolysis during chaperoning process. The gene coding for DNAJC14 protein is mapped to human chromosome 12q13.2.

Immunogen

DnaJ homolog subfamily C member 14 recombinant protein epitope signature tag (PrEST)

Application

Anti-DNAJC14 antibody produced in rabbit has been used for immunofluorescence studies and western blot analysis. All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DNAJC14 (DnaJ heat shock protein family (Hsp40) member C14) chaperone protein interacts with dopamine D1 receptor and regulates receptor trafficking between endoplasmic reticulum (ER) and plasma membrane. It also modulates the life cycles of various Flaviviridae family members and Flavivirus replication. DNAJC14 helps in SNARE (Soluble N-ethylmaleimide-sensitive factor activating protein receptor) complex-mediated transport by interacting with the lysosomal trafficking regulator protein.
DNAJC14 plays an important role in regulating yellow fever virus (YFV) replication complex assembly. The DNAJC family protein are also involved in various biological processes such as translation, exocytosis and endocytosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72886

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radiation hybrid mapping of equine CDK2, DGKA, DNAJC14, MMP19, CTSL and GAS1.
C Wittwer et al.
Animal genetics, 36(6), 536-537 (2005-11-19)
Yi-Qun Kuang et al.
PloS one, 7(7), e40522-e40522 (2012-07-21)
Chemokine receptors are members of the G protein-coupled receptor (GPCR) family. CCR5 and CXCR4 act as co-receptors for human immunodeficiency virus (HIV) and several efforts have been made to develop ligands to inhibit HIV infection by blocking those receptors. Removal
Zhigang Yi et al.
PLoS pathogens, 7(1), e1001255-e1001255 (2011-01-21)
Viruses in the Flavivirus genus of the Flaviviridae family are arthropod-transmitted and contribute to staggering numbers of human infections and significant deaths annually across the globe. To identify cellular factors with antiviral activity against flaviviruses, we screened a cDNA library
Zhigang Yi et al.
Journal of virology, 86(21), 11815-11832 (2012-08-24)
DNAJC14 is an Hsp40 family member that broadly modulates flavivirus replication. The mechanism by which DNAJC14 stoichiometrically participates in flavivirus replication complex (RC) formation is unknown; both reduced and elevated levels result in replication inhibition. Using yellow fever virus (YFV)
Leonia Bozzacco et al.
Journal of virology, 90(6), 3212-3228 (2016-01-08)
DNAJC14, a heat shock protein 40 (Hsp40) cochaperone, assists with Hsp70-mediated protein folding. Overexpressed DNAJC14 is targeted to sites of yellow fever virus (YFV) replication complex (RC) formation, where it interacts with viral nonstructural (NS) proteins and inhibits viral RNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service