Skip to Content
Merck
All Photos(5)

Key Documents

HPA015068

Sigma-Aldrich

Anti-CREB3L2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BBF2 human homolog on chromosome 7, Anti-cAMP-responsive element-binding protein 3-like protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

MEVLESGEQGVLQWDRKLSELSEPGDGEALMYHTHFSELLDEFSQNVLGQLLNDPFLSEKSVSMEVEPSPTSPAPLIQAEHSYSLCEEPRAQSPFTHITTSDSFND

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB3L2(64764)

General description

CREB3L2 (cAMP responsive element binding protein 3-like 2) is also called BBF2 human homolog on chromosome 7 (BBF2H7), and along with old astrocyte specifically induced substance (OASIS) is a member of the cAMP response element-binding protein (CREB)/ATF (Activating transcription factor) family. It is a transmembrane protein, and acts as a bZIP (basic leucine zipper) transcription factor. This gene has 12 exons, and covers >120kbp. It has a transactivation region in its N-terminal, a leucine zipper, a DNA-binding domain, a transmembrane α-helical hydrophobic region, and a luminal domain. It is an endoplasmic reticulum (ER) protein, but an alternatively spliced variant is found in both ER and nucleus.

Immunogen

cAMP-responsive element-binding protein 3-like protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-CREB3L2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CREB3L2 (cAMP responsive element binding protein 3-like 2) confers protection during endoplasmic reticulum (ER) stress, and in response to ER stress is cleaved in its transmembrane region. It regulates the transcription of Sec23A, when it acts like a bZIP transcription factor. This is essential for collagen synthesis in dermal fibroblasts, where it ensures the proper trafficking of procollagen from ER-to-Golgi. It is a fusion partner of FUS gene in low-grade fibromyxoid sarcoma (LGFMS), and this fusion forms a supernumerary ring chromosome, which is related to tumor progression. It is a substrate for F-box and WD repeat domain-containing-7 (Fbxw7) protein, and is degraded by the same. This process, in turn, regulates osteogenesis and chondrogenesis. It also acts as a fusion partner of PPAR (Peroxisome proliferator-activated receptor) γ in thyroid carcinoma, and this fusion inactivates a thyroid signaling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73339

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanae Yumimoto et al.
The Journal of biological chemistry, 288(40), 28488-28502 (2013-08-21)
Although identification of substrates for an enzyme is a key step in elucidation of its biological functions, detection of the interaction between enzymes and substrates remains challenging. We recently developed a new approach, termed differential proteomics-based identification of ubiquitylation substrates
Emely Möller et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(9), 2646-2656 (2011-05-04)
Low-grade fibromyxoid sarcoma (LGFMS) is typically characterized by the specific translocation t(7;16)(q33;p11) and the corresponding fusion gene FUS-CREB3L2. The present study aimed to extract LGFMS-specific, and putatively FUS-CREB3L2-dependent, gene expression patterns to learn more about the pathogenesis of this tumor.
Hammurabi Bartuma et al.
Cancer genetics and cytogenetics, 199(2), 143-146 (2010-05-18)
Low-grade fibromyxoid sarcoma (LGFMS) is a rare, low-grade malignant soft tissue tumor that is often mistaken for either benign or more malignant tumor types. Commonly, this tumor affects young adults and typically arises in the deep proximal extremities or trunk
Ioannis Panagopoulos et al.
Oncology reports, 24(5), 1133-1139 (2010-09-30)
CREB3L2, a member of the CREB3 family of transcription factors, spans >120 kbp and is composed of 12 exons. We characterized a widely expressed transcript of CREB3L2 generated by an intronic polyadenylation site in intron 4 of the gene. It
Weng-Onn Lui et al.
Cancer research, 68(17), 7156-7164 (2008-09-02)
The discovery of gene fusion mutations, particularly in leukemia, has consistently identified new cancer pathways and led to molecular diagnostic assays and molecular-targeted chemotherapies for cancer patients. Here, we report our discovery of a novel CREB3L2-PPARgamma fusion mutation in thyroid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service