Skip to Content
Merck
All Photos(4)

Key Documents

HPA014811

Sigma-Aldrich

Anti-GYPA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD235a antigen, Anti-Glycophorin-A precursor, Anti-MN sialoglycoprotein, Anti-PAS-2, Anti-Sialoglycoprotein alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

TSSSVTKSYISSQTNDTHKRDTYAATPRAHEVSEISVRTVYPPEEETGERVQLAHHFSEP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GYPA(2993)

General description

GYPA (glycophorin A) is the predominant sialoglycoprotein present in the human erythrocytes membrane. It carries the epitopes of blood groups M and N. It is a transmembrane protein, with a seven residue sequence L75IxxGVxxGVxxT87 present in its transmembrane helix. This sequence is essential for the dimerization of GYPA.

Immunogen

Glycophorin-A precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GYPA (glycophorin A) acts as a marker for intraplaque hemorrhage, and in coronary atheromas, is linked with the infiltration of macrophages and necrotic cores. It is up-regulated in symptomatic patients of carotid atherosclerotic lesions, as opposed to asymptomatic patients. Some portions of O-linked oligosaccharide chains of human GYPA contain blood group A, B or H antigens, which make up the ABO donor phenotype. In MN-heterozygous individuals, mutations in this gene are linked to the pathogenesis of chronic benzene poisoning.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72970

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puangpaka Chanpeng et al.
Scientific reports, 9(1), 6059-6059 (2019-04-17)
A hypercoagulable state leading to a high risk of a thrombotic event is one of the most common complications observed in β-thalassemia/HbE disease, particularly in patients who have undergone a splenectomy. However, the hypercoagulable state, as well as the molecular
Veerappan Anbazhagan et al.
Biochimica et biophysica acta, 1798(10), 1899-1907 (2010-07-07)
The influence of lipid bilayer properties on a defined and sequence-specific transmembrane helix-helix interaction is not well characterized yet. To study the potential impact of changing bilayer properties on a sequence-specific transmembrane helix-helix interaction, we have traced the association of
Cai-hong Xing et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 25(2), 87-90 (2007-04-26)
To explore the association of the glycophorin A(GPA) gene mutation in peripheral erythrocytes and chronic benzene poisoning. Sixty-three patients with chronic benzene poisonings and 45 benzene-exposed workers who were engaged in the same job title were investigated. Fluorescence immunolabeling technique
Sten-Ake Fredriksson et al.
Archives of biochemistry and biophysics, 498(2), 127-135 (2010-05-04)
We previously showed that a small proportion of the O-linked oligosaccharide chains of human glycophorin A (GPA) contains blood group A, B or H antigens, relevant to the ABO phenotype of the donor. The structures of these minor O-glycans have
M Hakimi et al.
International journal of molecular medicine, 32(2), 331-338 (2013-06-01)
The aim of this study was to evaluate in detail the histopathological characteristics of endarterectomized carotid atherosclerotic lesions in symptomatic versus asymptomatic patients. Twenty carotid lesions, 10 from asymptomatic and 10 from symptomatic patients who underwent carotid endarterectomy were classified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service