Skip to Content
Merck
All Photos(2)

Key Documents

HPA014738

Sigma-Aldrich

Anti-KCNF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Potassium voltage-gated channel subfamily F member 1, Anti-Voltage-gated potassium channel subunit Kv51, Anti-kH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

VRYYNKQRVLETAAKHELELMELNSSSGGEGKTGGSRSDLDNLPPEPAGKEAPSCSSRLKLSHSDTFIPLLTEEKHHRTRLQSCK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNF1(3754)

General description

KCNF1 (potassium channel, voltage gated modifier subfamily F, member 1) belongs to the family of voltage-gated Kv channels, which contains around 40 gene members. These channels are made of four subunits, and form heterotetramers with different members within this family. KCNF1 is also called Kv5.1, and acts as a modifier subunit, i.e. it is not functional on its own. It is expressed in brain, heart, liver, kidney, pancreas and skeletal muscles. This gene is localized to human chromosome 2p25, and codes for a protein which is composed of 495 amino acids.

Immunogen

Potassium voltage-gated channel subfamily F member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KCNF1 (potassium channel, voltage gated modifier subfamily F, member 1) is not functional in itself, but acts as a modifier channel for Kv2 family of channels. It regulates the functionality of Kv2.1 and Kv2.1 channels. It acts as the α-subunit of Kv2.1 channel, and increases the rate of inactivation, thereby increasing neuronal excitability and the duration of action potential.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72685

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Kramer et al.
The American journal of physiology, 274(6 Pt 1), C1501-C1510 (1998-06-05)
We have determined the effects of coexpression of Kv2.1 with electrically silent Kv5.1 or Kv6.1 alpha-subunits in Xenopus oocytes on channel gating. Kv2.1/5.1 selectively accelerated the rate ofinactivation at intermediate potentials (-30 to 0 mV), without affecting the rate at
Ching-Yi Chen et al.
Cancer gene therapy (2022-11-18)
Lung cancer continues to be the leading cause of cancer death in the United States. Despite recent advances, the five-year survival rate for lung cancer compared to other cancers still remains fairly low. The discovery of molecular targets for lung
K Su et al.
Biochemical and biophysical research communications, 241(3), 675-681 (1998-01-22)
Two novel human genes encoding putative potassium channels, kH1 and kH2, were identified from a human fetal brain cDNA library. Sequence analysis showed that kH1 and kH2 are homologous to rat IK8 and rat K13, respectively. The kH1 encodes a
International Union of Pharmacology. LIII. Nomenclature and molecular relationships of voltage-gated potassium channels.
George A Gutman et al.
Pharmacological reviews, 57(4), 473-508 (2005-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service