Skip to Content
Merck
All Photos(4)

Key Documents

HPA014060

Sigma-Aldrich

Anti-TMED2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Membrane protein p24A, Anti-Transmembrane emp24 domain-containing protein 2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

DAHAEECFFERVTSGTKMGLIFEVAEGGFLDIDVEITGPDNKGIYKGDRESSGKYTFAAHMDGTYKFCFSNRMSTMTPKIVMFTIDIGEAPKGQDMETEAHQNKLEEMINELAVAMTAVKHEQEY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMED2(10959)

General description

The gene TMED2 (transmembrane p24 trafficking protein 2) encodes a member of the transmembrane emp24 domain family of proteins that are involved in the transport of polypeptides from the endoplasmic reticulum and the Golgi. It is found to be expressed in syncytiotrophoblast, cytotrophoblast, and stromal cells in the human placenta. The gene is localized to human chromosome 12q24.31.

Immunogen

Transmembrane emp24 domain-containing protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene TMED2 (transmembrane p24 trafficking protein 2) encodes a type I transmembrane protein that is present in the Golgi apparatus. The N-terminal, also called the GOLD (Golgi dynamics) domain, of this protein interacts with the second extracellular loop of PAR-2 (Protease-activated receptor-2) and regulates its resensitization by influencing receptor trafficking. It participates in the maturation of calcium sensing receptor (CaSR) and prevents its degradation in the early secretory pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72826

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fortunato Morabito et al.
British journal of haematology, 146(1), 44-53 (2009-05-15)
IGHV mutational status and ZAP-70 or CD38 expression correlate with clinical course in B-cell chronic lymphocytic leukaemia (CLL). The three markers may be discordant in the single case and there is no consensus on their combined use in clinical practise.
Ann Stepanchick et al.
Biochemical and biophysical research communications, 395(1), 136-140 (2010-04-07)
The calcium sensing receptor (CaSR) is a Family 3/C G protein-coupled receptor with slow and partial targeting to the plasma membrane in both native and heterologous cells. We identified cargo receptor family member p24A in yeast two-hybrid screens with the
A Zakariyah et al.
Placenta, 33(3), 214-219 (2012-01-04)
Members of the transmembrane emp24 domain (Tmed)/p24 family of proteins are required for transport of proteins between the endoplasmic reticulum and the Golgi. One member of this family, Tmed2/p24β1, is expressed during placental development in mice and its expression is
Weibo Luo et al.
The Journal of biological chemistry, 282(41), 30246-30255 (2007-08-19)
Protease-activated receptor-2 (PAR-2), the second member of the G protein-coupled PAR family, is irreversibly activated by trypsin or tryptase and then targeted to lysosomes for degradation. Intracellular presynthesized receptors stored at the Golgi apparatus repopulate the cell surface after trypsin
Gong Shi-Peng et al.
Oncotarget, 8(55), 94151-94165 (2017-12-08)
TMED2 is involved in morphogenesis of the mouse embryo and placenta. We found that expression of TMED2 was higher in epithelial ovarian cancer tissues than normal ovarian tissues. Silencing TMED2 decreased cell proliferation, migration, and invasion. Ectopic expression of TMED2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service