Skip to Content
Merck
All Photos(7)

Key Documents

HPA011216

Sigma-Aldrich

Anti-CD207 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-type lectin domain family 4 member K antibody produced in rabbit, Anti-CD207 antigen antibody produced in rabbit, Anti-Langerin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VKTNVQLLKGRVDNISTLDSEIKKNSDGMEAAGVQIQMVNESLGYVRSQFLKLKTSVEKANAEIQILTRSWEEVSTLNAQIPELKSDLEKASALNTKIRALQGSLENMSKLLKRQNDILQVVSQGWKYFKGNFYYFSLIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD207(50489)

General description

Langerin (CD207) is a C-type lectin, which is expressed on Langerhans cells. It contains a C-type carbohydrate recognition domain (CRD) in its extracellular region. It also contains a Ca2+-dependent sugar binding region. It is a type II transmembrane protein, and its extracellular region also contains the neck region. This neck region forms coiled-coils of α-helices which give rise to oligomers of CD207. It contains a proline-rich motif (WPREPPP) in its intracellular domain, which in turn is composed of 43 amino acids. This proline-rich motif might be involved in signal transduction. CD207 gene is localized to human chromosome 2p13.

Immunogen

C-type lectin domain family 4 member K recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Langerin (CD207) is an endocytic receptor which is involved in the formation of Birbeck granules, which are a part of the endosomal compartments exclusive to Langerhans cells. It recognizes the surface glucoconjugates present on pathogens such as, human immunodeficiency virus (HIV), and internalizes them into Birbeck granules. It therefore, prevents the transfer of HIV from Langerhans cells to T-cells, by degrading it. It forms the first step in antigen processing and presentation to immune cells. It recognizes a wide range of pathogens such as, Mycobacterium leprae, herpes simplex virus 2, and fungi such as, Malassezia furfur and Candida species. CD207 also binds to measles virus and presents it to CD4+ T cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71827

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hadar Feinberg et al.
The Journal of biological chemistry, 288(52), 36762-36771 (2013-11-13)
Langerin, a C-type lectin on Langerhans cells, mediates carbohydrate-dependent uptake of pathogens in the first step of antigen presentation to the adaptive immune system. Langerin binds a diverse range of carbohydrates including high mannose structures, fucosylated blood group antigens, and
J Valladeau et al.
Immunity, 12(1), 71-81 (2000-02-08)
We have identified a type II Ca2+-dependent lectin displaying mannose-binding specificity, exclusively expressed by Langerhans cells (LC), and named Langerin. LC are uniquely characterized by Birbeck granules (BG), which are organelles consisting of superimposed and zippered membranes. Here, we have
Michiel van der Vlist et al.
European journal of immunology, 41(9), 2619-2631 (2011-07-09)
Langerhans cells (LCs) are a subset of DCs that reside in the upper respiratory tract and are ideally suited to sense respiratory virus infections. Measles virus (MV) is a highly infectious lymphotropic and myelotropic virus that enters the host via
Hadar Feinberg et al.
The Journal of biological chemistry, 285(17), 13285-13293 (2010-02-26)
Langerin, an endocytic receptor of Langerhans cells, binds pathogens such as human immunodeficiency virus by recognition of surface glycoconjugates and mediates their internalization into Birbeck granules. Langerin has an extracellular region consisting of a C-type carbohydrate-recognition domain (CRD) and a
Małgorzata Anna Cichoń et al.
eLife, 11 (2022-10-13)
Skin is an active immune organ where professional antigen-presenting cells such as epidermal Langerhans cells (LCs) link innate and adaptive immune responses. While Reticulon 1A (RTN1A) was recently identified in LCs and dendritic cells in cutaneous and lymphoid tissues of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service