Skip to Content
Merck
All Photos(7)

Key Documents

HPA010926

Sigma-Aldrich

Anti-ALCAM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Activated leukocyte-cell adhesion molecule antibody produced in rabbit, Anti-CD166 antigen precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

NITLKCLGNGNPPPEEFLFYLPGQPEGIRSSNTYTLTDVRRNATGDYKCSLIDKKSMIASTAITVHYLDLSLNPSGEVTRQIGDALPVSCTISASRNATVVWMKDNIRLRSSPSFSSLHYQDAGNYVCETALQEVEGLKKR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALCAM(214)

General description

Activated leukocyte cell adhesion molecule (ALCAM) belongs to the superfamily of immunoglobulin cell adhesion molecules (Ig-CAMs). Its extracellular domain consist of five immunoglobulin (Ig)-like domains, which aid in cell-cell adhesion, either through heterophilic (ALCAM-CD6) or homophilic (ALCAM-ALCAM) interactions. It is a type I transmembrane protein. It has a short cytoplasmic tail and a transmembrane region, and has a molecular weight of 110kDa. It is expressed in a wide range of tissues, especially in the epithelia and the corresponding cancers. ALCAM gene maps to human chromosome 3q13.1-q13.2.

Immunogen

CD166 antigen precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ALCAM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Activated leukocyte cell adhesion molecule (ALCAM) mediates cell-cell adhesion, either through heterophilic (ALCAM-CD6) or homophilic (ALCAM-ALCAM) interactions. Hemophilic interactions aid in cell migration and invasiveness of tumor cells. ALCAM present on dendritic cells, induces T-cell activation by interacting with cluster of differentiation 6 (CD6) present on T-cells. This heterophilic interaction is necessary for the stabilization of immunological synapses. It mediates the extravasation of leukocytes and their transport to the central nervous system, as well as axonal fasciculation. Colorectal cancer patients, who show increased levels of ALCAM shedding, have poor prognosis and survival rates. The soluble form of ALCAM is related to the aggressive type II phenotype of epithelial ovarian cancer, and can be used as a marker for the same.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72229

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ALCAM: Basis Sequence: Mouse.
Amanda G Hansen et al.
The AFCS-nature molecule pages, 2011 (2011-01-01)
Pedro Nicolau-Neto et al.
Cancers, 12(2) (2020-02-23)
Laryngeal squamous cell carcinoma (LSCC) is one of the most incident tumors in the world, especially in developing countries, such as Brazil. Different from other tumors, LSCC prognosis did not improve during the past four decades. Therefore, the objective of
Alvaro Gilsanz et al.
Cellular and molecular life sciences : CMLS, 70(3), 475-493 (2012-10-12)
ALCAM/CD166 is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs) which mediates intercellular adhesion through either homophilic (ALCAM-ALCAM) or heterophilic (ALCAM-CD6) interactions. ALCAM-mediated adhesion is crucial in different physiological and pathological phenomena, with particular relevance in leukocyte
Svitlana Tyekucheva et al.
Nature communications, 8(1), 420-420 (2017-09-06)
While progression from normal prostatic epithelium to invasive cancer is driven by molecular alterations, tumor cells and cells in the cancer microenvironment are co-dependent and co-evolve. Few human studies to date have focused on stroma. Here, we performed gene expression
Joost Te Riet et al.
Journal of cell science, 127(Pt 7), 1595-1606 (2014-02-06)
At the immunological synapse, the activated leukocyte cell adhesion molecule (ALCAM) on a dendritic cell (DC) and CD6 molecules on a T cell contribute to sustained DC-T-cell contacts. However, little is known about how ALCAM-CD6 bonds resist and adapt to

Articles

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service