Skip to Content
Merck
All Photos(4)

Key Documents

HPA010718

Sigma-Aldrich

Anti-FCGR2A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD32 antigen antibody produced in rabbit, Anti-CDw32 antibody produced in rabbit, Anti-Fc-γ RII-a antibody produced in rabbit, Anti-Fc-γ-RIIa antibody produced in rabbit, Anti-FcRII-a antibody produced in rabbit, Anti-IgG Fc receptor II-a antibody produced in rabbit, Anti-Low affinity immunoglobulin γ Fc region receptor II-a precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RISANSTDPVKAAQFEPPGRQMIAIRKRQLEETNNDYETADGGYMTLNPRAPTDDDKNIYLTLPPNDHVNSNN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCGR2A(2212)

Immunogen

Low affinity immunoglobulin γ Fc region receptor II-a precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FCGR2A antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FCGR2A (Fc fragment of IgG, low affinity IIa, receptor) gene encodes a low affinity receptor belonging to the family of immunoglobulin Fc receptors that are found on the surface of many immune response cells. The encoded receptor is expressed on both myeloid and lymphoid cells. It plays an important role in phagocytosis, antibody-dependent cellular cytotoxicity (ADCC), and the release of mediators of inflammation. It binds to IgG and triggers cellular responses against pathogens and soluble antigens. The receptor is also involved in the regulation of differentiation of lymphocytes and the production of antibodies. This gene has been identified as a susceptibility locus for Kawasaki disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72030

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison Ruth Jennings et al.
Brain pathology (Zurich, Switzerland), 25(5), 517-530 (2014-09-02)
Reports that chronically demyelinated multiple sclerosis brain and spinal cord lesions contained immature oligodendrocyte lineage cells have generated major interest aimed at the potential for promotion of endogenous repair. Despite the prominence of the optic nerve as a lesion site
Anthony J Smith et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 3126-3133 (2014-08-22)
Principles to guide design of an effective vaccine against HIV are greatly needed, particularly to protect women in the pandemic's epicenter in Africa. We have been seeking these principles by identifying correlates of the robust protection associated with SIVmac239Δnef vaccination
D G Brooks et al.
The Journal of experimental medicine, 170(4), 1369-1385 (1989-10-01)
The structural heterogeneity of the human low affinity receptor for IgG, FcRII(CD32), has been elucidated through the isolation, characterization, and expression of cDNA clones derived from myeloid and lymphoid RNA. These clones predict amino acid sequences consistent with integral membrane
Chiea Chuen Khor et al.
Nature genetics, 43(12), 1241-1246 (2011-11-15)
Kawasaki disease is a systemic vasculitis of unknown etiology, with clinical observations suggesting a substantial genetic contribution to disease susceptibility. We conducted a genome-wide association study and replication analysis in 2,173 individuals with Kawasaki disease and 9,383 controls from five
Jinghua Lu et al.
Nature, 456(7224), 989-992 (2008-11-18)
Pentraxins are a family of ancient innate immune mediators conserved throughout evolution. The classical pentraxins include serum amyloid P component (SAP) and C-reactive protein, which are two of the acute-phase proteins synthesized in response to infection. Both recognize microbial pathogens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service