Skip to Content
Merck
All Photos(3)

Key Documents

HPA010024

Sigma-Aldrich

Anti-CADM2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell adhesion molecule 2 precursor, Anti-Immunoglobulin superfamily member 4D, Anti-Nectin-like protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

EIHYTPSVKIIPSTPFPQEGQPLILTCESKGKPLPEPVLWTKDGGELPDPDRMVVSGRELNILFLNKTDNGTYRCEATNTIGQSSAEYVLIVHDVPNTLLPTTIIPSLTTATVTTTVAITTSPTTSATTSSIRDPNALA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CADM2(253559)

General description

CADM2 (cell adhesion molecule 2) is a member of the CADM family, which in turn belongs to the immunoglobulin (Ig) superfamily. All the members of this family are composed of three extracellular Ig-like loops, a transmembrane domain and an intracellular region. This gene is localized to human chromosome 3p12.1, and is alternatively spliced to two isoforms- CADM2a and CADM2b.

Immunogen

Cell adhesion molecule 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CADM2 (cell adhesion molecule 2) is significantly donw-regulated in HCC (hepatocellular carcinoma), and this is linked with poor recurrence-free survival (RFS). Its expression in HCC is linked with differentiation, serum a-fetoprotein, vascular invasion and hepatitis B surface antigen (HBsAg). CADM2 functions as a tumor suppressor, and abnormal methylation leading to its inactivation is linked with human renal cell carcinoma tumor progression and human prostate cancer. It functions as a guidance molecule for post-crossing commissural axons, and is critical for the formation and plasticity of synapse. Inactivation of this gene results in abnormal axon-axon interaction, leading to deregulated pathfinding of sensory neurons. A new splicing variant of CADM2 is a pathogenesis factor for psoriasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71600

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sen Yang et al.
Journal of cancer research and clinical oncology, 140(1), 109-116 (2013-11-19)
To investigate the expression and clinical significance of CADM2 in hepatocellular carcinomas (HCC). The level of expression of CADM2 mRNA was assessed in frozen tumor specimens and adjacent noncancerous tissues from 30 HCC patients by real-time PCR. The protein level
Guimin Chang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(22), 5390-5401 (2010-11-11)
Cell adhesion molecules (CADM) comprise a newly identified protein family whose functions include cell polarity maintenance and tumor suppression. CADM-1, CADM-3, and CADM-4 have been shown to act as tumor suppressor genes in multiple cancers including prostate cancer. However, CADM-2
Wei He et al.
Biochemical and biophysical research communications, 435(4), 526-532 (2013-05-07)
Cell adhesion molecules (CADMs) comprise a protein family whose functions include maintenance of cell polarity and tumor suppression. In this report, we show that the CADM2 gene is repressed in human clear renal cell carcinoma by DNA promoter hypermethylation and/or
Azusa Hiruma et al.
Biochemical and biophysical research communications, 412(4), 626-632 (2011-08-26)
CADM2, a candidate gene for psoriasis, was identified by a genome-wide association study using microsatellites in the Japanese population (561 cases and 561 controls). Moreover, haplotype analysis included an additional 68 SNPs and indicated that a 110-kb haplotype block was
Jeannine A Frei et al.
Journal of cell science, 127(Pt 24), 5288-5302 (2014-10-23)
Synaptic cell adhesion molecules (SynCAMs) are crucial for synapse formation and plasticity. However, we have previously demonstrated that SynCAMs are also required during earlier stages of neural circuit formation because SynCAM1 and SynCAM2 (also known as CADM1 and CADM2, respectively)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service