Skip to Content
Merck
All Photos(3)

Key Documents

HPA008832

Sigma-Aldrich

Anti-NFASC antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neurofascin precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LECRVKHDPSLKLTVSWLKDDEPLYIGNRMKKEDDSLTIFGVAERDQGSYTCVASTELDQDLAKAYLTVLADQATPTNRLAALPKGRPDRPRDLELTDLAERSVRLTWIPGDANNSPITDYVVQFE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFASC(23114)

General description

NFASC (neurofascin) is a member of the L1 family of CAMs (cell adhesion molecules), which in turn is a part of the immunoglobulin (Ig) superfamily of proteins. It is mainly expressed in the nervous system. The members of L1 family contain six Ig-like domains and four to five Fn (fibronectin) domains in their extracellular region. Their cytoplasmic domain contains a putative conserved motif (FIG(Q/A)Y) that links the proteins to cytoplasm and also facilitates the binding to ankyrin. This protein has multiple spliced isoforms, with two major isoforms called NF155 of 155kDa and NF186 of 186kDa. These isoforms are expressed in the glia and axons respectively.

Immunogen

Neurofascin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The NF155 isoform of NFASC (neurofascin) acts as the ligand for CNTN1/CASPR1 (contactin 1/ contactin-associated protein 1) complex at the paranode of glia, where it plays an essential role in myelination and the functional assembly of node of Ranvier. Inactivation of this isoform in mice results in severe demyelinating neuropathy, with highly decreased conduction velocities. It is also accompanied with degeneration of Purkinje neurons, which results in ataxia and prominent tremor. Autoantibodies to this protein are linked to a subtype of chronic inflammatory demyelinating polyradiculoneuropathy (CIDP). Studies show that antibodies against NFASC result in accumulation of complement proteins, axonal injury and the exacerbation of multiple sclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71582

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heli Liu et al.
The Journal of biological chemistry, 286(1), 797-805 (2010-11-05)
The L1 family neural cell adhesion molecules play key roles in specifying the formation and remodeling of the neural network, but their homophilic interaction that mediates adhesion is not well understood. We report two crystal structures of a dimeric form
Yuji Nishimura et al.
Brain pathology (Zurich, Switzerland), 33(3), e13131-e13131 (2022-11-12)
The pathological hallmark of multiple system atrophy (MSA) is aberrant accumulation of phosphorylated α-synuclein in oligodendrocytes, forming glial cytoplasmic inclusions (GCIs). Extensive demyelination occurs particularly in the olivopontocerebellar and striatonigral pathways, but its precise mechanism remains elusive. Glial connexins (Cxs)
Luis Querol et al.
Neurology, 82(10), 879-886 (2014-02-14)
To describe the frequency of antibodies against neurofascin in chronic inflammatory demyelinating polyradiculoneuropathy (CIDP) and the associated clinical features. Immunocytochemistry was used to identify antibodies to neurofascin 155 (NF155) and 186. Serum reactivity with paranodes and brain tissue was tested
Emily K Mathey et al.
The Journal of experimental medicine, 204(10), 2363-2372 (2007-09-12)
Axonal injury is considered the major cause of disability in patients with multiple sclerosis (MS), but the underlying effector mechanisms are poorly understood. Starting with a proteomics-based approach, we identified neurofascin-specific autoantibodies in patients with MS. These autoantibodies recognize the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service