Skip to Content
Merck
All Photos(4)

Key Documents

HPA007875

Sigma-Aldrich

Anti-MMP3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-3 antibody produced in rabbit, Anti-Matrix metalloproteinase-3 antibody produced in rabbit, Anti-SL-1 antibody produced in rabbit, Anti-Stromelysin-1 precursor antibody produced in rabbit, Anti-Transin-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MYPLYHSLTDLTRFRLSQDDINGIQSLYGPPPDSPETPLVPTEPVPPEPGTPANCDPALSFDAVSTLRGEILIFKDRHFWRKSLRKLEPELHLISSFWPSLPSGVDAAYEVTSKDLVFIFKGNQFWAI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP3(4314)

General description

MMP3 (matrix metallopeptidase 3) gene is a part of a gene cluster that encodes matrix metalloproteinases (MMP) and is localized to human chromosome 11q22.3. MMPs are a family of 23 members that are induced by the cytokine tumor necrosis factor (TNF)-α. These proteinases are secreted in an inactive form and are activated upon cleavage by certain proteinases.

Immunogen

Stromelysin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-MMP3 antibody produced in rabbit has been used for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-MMP3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Matrix metallopeptidase 3 (MMP3) has a broad substrate specificity and is capable of degrading fibronectin, laminin, collagens III, IV, IX, and X, and cartilage proteoglycans. This enzyme functions is several physiological processes, such as in wound repair, progression of atherosclerosis, tissue remodeling and tumor initiation. MMP3 variant can be associated with hypertrophy of interventricular septum or hypertrophic cardiomyopathy. It may serves as a non-invasive biomarker of histological synovitis and diagnosis of rheumatoid arthritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70114

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Da Ma et al.
Mediators of inflammation, 2014, 179284-179284 (2014-08-26)
To explore the correlation between matrix metalloproteinase- (MMP-) 3 and histological synovitis in rheumatoid arthritis (RA). Serum MMP-3 of 62 patients with active RA was detected by ELISA. Serial synovial tissue sections from all RA patients, 13 osteoarthritis, and 10
David Tong et al.
Heliyon, 10(13), e32891-e32891 (2024-07-19)
Regenerative endodontic procedures (REP) aim at reestablishing tooth vitality by replacing the irreversibly damaged dental pulp removed by the dental practitioner with a new functional one. The current treatment of advanced caries relies on the replacement of the inflamed or
E V Privalova et al.
Kardiologiia, 54(5), 4-7 (2014-09-02)
Prognosis of patients with hypertrophic cardiomyopathy (HCMP) to a great extent is determined by clinical variant of the disease. As the system of matrix metalloproteinases (MMPs) plays an important role in development and progression of the processes of fibroformation and
Jun-Jie Chen et al.
Asian Pacific journal of tropical medicine, 7(4), 297-300 (2014-02-11)
To investigate the expression and significance of MMP-3 in synovium of knee joint at different stage in osteoarthritis (OA) patients. Knee synovial tissue were collected in 90 OA patients (the OA group). Patients in the OA group was divided into
Magnus S Ågren et al.
European journal of cell biology, 94(1), 12-21 (2014-12-03)
Tumor necrosis factor (TNF)-α induces matrix metalloproteinases (MMPs) that may disrupt skin integrity. We have investigated the effects and mechanisms of exogenous TNF-α on collagen degradation by incubating human skin explants in defined serum-free media with or without TNF-α (10ng/ml)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service