Skip to Content
Merck
All Photos(3)

Key Documents

HPA006411

Sigma-Aldrich

Anti-MUC7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apo-MG2, Anti-MUC-7, Anti-Mucin-7 precursor, Anti-Salivary mucin-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RRHHHQSPKSHFELPHYPGLLAHQKPFIRKSYKCLHKRCRPKLPPSPNNPPKFPNPHQPPKHPDKNSSVVNPTLVATTQIPSVTFPSASTKITTLPNVTFLPQNATTISSRENVNTSSSVATLAPVNSPAP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC7(4589)

General description

MUC7 (mucin 7) is the lower molecular weight (200- 300kDa) mucin of the two types of oral cavity mucins, and is also called MG2. The other type of mucin, MG1, has high molecular weight (1000kDa). This gene is localized to human chromosome 4q13-q21, and is highly polymorphic in nature. This protein is composed of three domains namely, 4 and 1 potential N- glycosylation sites at its either ends, and a tandem repeat domain located centrally. MUC7 is composed of 68% carbohydrate and 30% protein. This protein has a molecular weight of 180kDa, and is highly expressed in sublingual and submandibular glands.

Immunogen

Mucin-7 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MUC7 (mucin 7) has anti-bacterial, anti-viral and anti-fungal properties. It is expressed in the airway secretions of asthmatics. Variants in this gene are linked to decreased susceptibility to asthma in African-American population. In patients with rheumatoid arthritis, this protein is highly sulfated. MUC7*6/*6 polymorphism is linked to higher oral hygiene, as well as higher chances of dental caries.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70011

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah A Flowers et al.
Molecular & cellular proteomics : MCP, 12(4), 921-931 (2013-03-05)
Rheumatoid arthritis is a common and debilitating systemic inflammatory condition affecting up to 1% of the world's population. This study aimed to investigate the immunological significance of O-glycans in chronic arthritis at a local and systemic level. O-Glycans released from
Kristina A Thomsson et al.
Methods in molecular biology (Clifton, N.J.), 742, 127-141 (2011-05-07)
The major phenotype of CF is the accumulation of mucus, a phenomenon whose relation to the dysfunctional CFTR is still not fully understood. This means that studies of mucus and its main component, the mucins, are important. Due to the
Jadwiga Buczkowska-Radlińska et al.
Postepy higieny i medycyny doswiadczalnej (Online), 66, 204-209 (2012-06-19)
The aim of the study was to analyze polymorphism of the MUC7 gene and its correlation with the DMFT value and the Plaque Control Record by O'Leary. The study was carried out on 158 students of a faculty of dentistry
Nayab M A Chaudhury et al.
Molecular & cellular proteomics : MCP, 15(3), 1048-1059 (2015-12-04)
Sjögren's syndrome is a chronic autoimmune disorder characterized by lymphocytic infiltration and hypofunction of salivary and lacrimal glands. This loss of salivary function leads to oral dryness, impaired swallowing and speech, and increased infection and is associated with other autoimmune
Justyna Pol
Annales Academiae Medicae Stetinensis, 57(2), 85-91 (2011-01-01)
The aim of this study was to search for an association of the polymorphism of MUC7 gene encoding the low-molecular-weight mucin MG2 with susceptibility to caries. The study enrolled 158 students (age 20-21 years) of the first and second year

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service