Skip to Content
Merck
All Photos(5)

Key Documents

HPA005714

Sigma-Aldrich

Anti-FOXJ1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

FOXJ1 Antibody - Anti-FOXJ1 antibody produced in rabbit, Foxj1 Antibody, Anti-Forkhead box protein J1 antibody produced in rabbit, Anti-Forkhead-related protein FKHL13 antibody produced in rabbit, Anti-HFH-4 antibody produced in rabbit, Anti-Hepatocyte nuclear factor 3 forkhead homolog 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

PREKDEPGKGGFWRIDPQYAERLLSGAFKKRRLPPVHIHPAFARQAAQEPSAVPRAGPLTVNTEAQQLLREFEEATGEAGWGAGEGRLGHKRKQPLPKRVAKVPRPPSTLLPTPEEQGELEPLKG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXJ1(2302)

General description

Forkhead box protein J1 (FOXJ1) is a transcription factor which belongs to the forkhead-box (FOX) gene family. It contains a DNA-binding domain called the forkhead domain. The gene encoding the protein is present on chromosome 17q25.

Immunogen

Forkhead box protein J1 recombinant protein epitope signature tag (PrEST)

Application

Anti-FOXJ1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Forkhead box protein J1 (FOXJ1) is a transcription factor that can suppress T cell activity, through the repression of NF-κβ activity. Its dysregulation is associated with autoimmune diseases and inflammatory diseases. FOXJ1 plays a critical role in the maintenance of immune homeostasis. This involves intricate interactions between the forkhead family members and inflammatory transcription factors. It coordinates the regulation of the activity of two key inflammatory transcription factors, NF-AT and NF-κβ. It has also been shown that FOXJ1 has fundamental roles in ciliogenesis in the lung and is directly required for epithelial cell ciliogenesis of the neonatal oviduct.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86948

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Peng et al.
Allergy, asthma, and clinical immunology : official journal of the Canadian Society of Allergy and Clinical Immunology, 14, 71-71 (2018-11-22)
Upper airway inflammatory diseases are associated with abnormal expression of nasal epithelial forkhead-box J1 (FOXJ1) which regulates motile cilia formation. We sought to investigate whether aberrant FOXJ1 localizations correlate with the disease severity and the co-existence of allergic rhinitis (AR)
Yang Peng et al.
International archives of allergy and immunology, 176(2), 115-123 (2018-04-11)
Forkhead box J1 (FOXJ1) plays pivotal roles in motile cilia formation. However, it remains unclear whether abnormal expression or localization of FOXJ1 in nasal mucosa tissues is associated with allergic rhinitis (AR), in which impaired mucociliary clearance is implicated. We
Identification of single nucleotide polymorphisms in FOXJ1 and their association with allergic rhinitis.
CS Li
Journal of Human Genetics, 51(4), 292-297 (2006)
Yue-Ying Yang et al.
Journal of inflammation research, 15, 3661-3675 (2022-07-06)
Radiotherapy (RT) is the mainstay treatment for head and neck cancers. However, chronic and recurrent upper respiratory tract infections and inflammation have been commonly reported in patients post-RT. The underlying mechanisms remain poorly understood. We used a well-established model of
Coordination of NF-kappaB and NFAT antagonism by the forkhead transcription factor Foxd1.
L Lin and SL Peng
Journal of Immunology, 176(8), 4793-4803 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service