Skip to Content
Merck
All Photos(2)

Key Documents

HPA005495

Sigma-Aldrich

Anti-GYG2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GN-2 antibody produced in rabbit, Anti-GN2 antibody produced in rabbit, Anti-Glycogenin-2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

PAFKQFGSSAKVVHFLGSMKPWNYKYNPQSGSVLEQGSASSSQHQAAFLHLWWTVYQNNVLPLYKSVQAGEARASPGHTLCHSDVGGPCADSASGVGEPCENSTPSAGVPCANSPLGSNQPAQGLPEPTQIVDETLSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GYG2(8908)

General description

Glycogenin 2 (GYG2) is a key enzyme in the synthesis of glycogen. It is a self-glycosylating protein. This gene spans 46kb, has 11 exons and localizes to chromosome Xp22.3. GYG2 is expressed in liver, heart and pancreas. It has 3 isoforms- glycogenins-α, β and γ. Glycogenin-2β is the major isoform to be expressed in liver.

Immunogen

Glycogenin-2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glycogenin-2 (GYG2) plays a major role in the synthesis of glycogen in liver. It initiates glycogen synthesis by forming the first glucose-O-tyrosine linkage in the presence of UDP-glucose. It forms homodimers as well as heterodimers with glycogenin-1. Glycogenin-1 facilitates self-glucosylation of glycogenin-2, with the vice-versa not being true. GYG2 can regulate glycogen synthesis, and the levels of GYG2 are correlated to the amount of glycogen present in liver. Studies suggest that there might be an association between mutations in this gene and Leigh syndrome (LS), which is an early-onset neurodegenerative disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74336

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Nilsson et al.
Biochimica et biophysica acta, 1844(2), 398-405 (2013-11-19)
Glycogen synthesis is initiated by self-glucosylation of the glycosyltransferases glycogenin-1 and -2 that, in the presence of UDP-glucose, form both the first glucose-O-tyrosine linkage, and then stepwise add a series of α1,4-linked glucoses to a growing chain of variable length.
Eri Imagawa et al.
Human genetics, 133(2), 225-234 (2013-10-09)
Leigh syndrome (LS) is an early-onset progressive neurodegenerative disorder characterized by unique, bilateral neuropathological findings in brainstem, basal ganglia, cerebellum and spinal cord. LS is genetically heterogeneous, with the majority of the causative genes affecting mitochondrial malfunction, and many cases
J Mu et al.
The Journal of biological chemistry, 273(52), 34850-34856 (1998-12-18)
Glycogenin-2 is a recently described self-glucosylating protein potentially involved in the initiation of glycogen biosynthesis (Mu, J., Skurat, A. V., and Roach, P. J. (1997) J. Biol. Chem. 272, 27589-27597). In human liver extracts, most of the glycogenin-2 was only
J Mu et al.
The Journal of biological chemistry, 272(44), 27589-27597 (1997-11-05)
Glycogenin is a self-glucosylating protein involved in the initiation phase of glycogen biosynthesis. A single mammalian gene had been reported to account for glycogen biogenesis in liver and muscle, the two major repositories of glycogen. We describe the characterization of
L Zhai et al.
Gene, 242(1-2), 229-235 (2000-03-18)
Glycogenin-2 is one of two self-glucosylating proteins involved in the initiation phase of the synthesis of the storage polysaccharide glycogen. Cloning of the human glycogenin-2 gene, GYG2, has revealed the presence of 11 exons and a gene of more than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service