Skip to Content
Merck
All Photos(4)

Key Documents

HPA005438

Sigma-Aldrich

Anti-HNF4G antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HNF-4-gamma, Anti-Hepatocyte nuclear factor 4-gamma, Anti-Nuclear receptor subfamily 2 group A member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000-1:2500

immunogen sequence

WQMIEQIQFVKLFGMVKIDNLLQEMLLGGASNDGSHLHHPMHPHLSQDPLTGQTILLGPMSTLVHADQISTPETPLPSPPQGSGQEQYKIAANQASVISHQHLSKQKQ

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF4G(3174)

General description

Hepatocyte nuclear factor -4 γ (HNF4G) belongs to the nuclear receptor superfamily and is an orphan receptor with significant homology to HNF4α. The gene for HNF-4γ is located on chromosome 8q. HNF-4γ is expressed in small intestine, testis, pancreas and kidney. Studies reveal that the ligand binding domain of HNF-4γ has constitutively bound fatty acids, which are endogenous in origin. HNF-4γ protein is composed of 774 amino acids.

Immunogen

Hepatocyte nuclear factor 4-gamma recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Hepatocyte nuclear factor -4 γ (HNF4G) protein forms heterodimers with HNF-4α, and together they are involved in the regulation of metabolism. They also induce the expression of various genes, including cell death-inducing DEF45-like effector b (CIDEB), which regulates lipid metabolism in the liver. In humans, the expression of dihydrodiol dehydrogenase (DD) is regulated by HNF-4γ. DD is responsible for the reduction of various ketone-containing drugs and the oxidation of trans-dihydrodiols of polycyclic aromatic hydrocarbons.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70753

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Plengvidhya et al.
Diabetes, 48(10), 2099-2102 (1999-10-08)
The aim of this study was to investigate whether mutations in hepatocyte nuclear factor (HNF)-4gamma, a transcription factor homologous to HNF-4alpha, contribute to the etiology of early-onset type 2 diabetes. Linkage between diabetes and two polymorphic markers at the HNF-4gamma
Takeshi Ozeki et al.
Pharmacogenetics, 13(1), 49-53 (2003-01-25)
Human dihydrodiol dehydrogenase (DD) catalyses the oxidation of trans-dihydrodiols of polycyclic aromatic hydrocarbons and the reduction of several ketone-containing drugs. About 40-fold interindividual difference in DD activities has been noted. Recently, we found that transcriptional factors, hepatocyte nuclear factor (HNF)-1
G Bruce Wisely et al.
Structure (London, England : 1993), 10(9), 1225-1234 (2002-09-11)
The 2.7 A X-ray crystal structure of the HNF4gamma ligand binding domain (LBD) revealed the presence of a fatty acid within the pocket, with the AF2 helix in a conformation characteristic of a transcriptionally active nuclear receptor. GC/MS and NMR
Rik Gh Lindeboom et al.
Molecular systems biology, 14(6), e8227-e8227 (2018-06-28)
Intestinal organoids accurately recapitulate epithelial homeostasis in vivo, thereby representing a powerful in vitro system to investigate lineage specification and cellular differentiation. Here, we applied a multi-omics framework on stem cell-enriched and stem cell-depleted mouse intestinal organoids to obtain a holistic view
T Drewes et al.
Molecular and cellular biology, 16(3), 925-931 (1996-03-01)
Hepatocyte nuclear factor 4 (HNF4) was first identified as a DNA binding activity in rat liver nuclear extracts. Protein purification had then led to the cDNA cloning of rat HNF4, which was found to be an orphan member of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service