Skip to Content
Merck
All Photos(9)

Key Documents

HPA004689

Sigma-Aldrich

Anti-ARHGAP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ρ GTPase-activating protein 1 antibody produced in rabbit, Anti-ρ-Related small GTPase protein activator antibody produced in rabbit, Anti-CDC42 GTPase-activating protein antibody produced in rabbit, Anti-GTPase-activating protein rhoOGAP antibody produced in rabbit, Anti-p50-RhoGAP antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SEALNQLKLASIDEKNWPSDEMPDFPKSDDSKSSSPELVTHLKWDDPYYDIARHQIVEVAGDDKYGRKIIVFSACRMPPSHQLDHSKLLGYLKHTLDQYVESDYTLLYLHHGLTSDNKPSLS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGAP1(392)

Looking for similar products? Visit Product Comparison Guide

Immunogen

ρ GTPase-activating protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-ARHGAP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ARHGAP1 is a Rho-GTPase activating protein. It is mainly involved in the Rho-mediated signaling pathway. In addition, it is also associated with other biological functions including cytoskeleton formation, JNK (c-Jun N-terminal kinase) signaling pathway and cell proliferation. It functions as a regulator switch between switch I and II regions i.e. active GTP-bound form and the inactive GDP-bound state. Generally, hydrolytic activity of the protein is low but interaction with rhoGAP can increase the hydrolysis of GTP to GDP.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86872

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Rittinger et al.
Nature, 389(6652), 758-762 (1997-10-24)
Small G proteins of the Rho family, which includes Rho, Rac and Cdc42Hs, regulate phosphorylation pathways that control a range of biological functions including cytoskeleton formation and cell proliferation. They operate as molecular switches, cycling between the biologically active GTP-bound
K Rittinger et al.
Nature, 388(6643), 693-697 (1997-08-14)
Small G proteins transduce signals from plasma-membrane receptors to control a wide range of cellular functions. These proteins are clustered into distinct families but all act as molecular switches, active in their GTP-bound form but inactive when GDP-bound. The Rho
V Härmä et al.
Oncogene, 31(16), 2075-2089 (2011-10-15)
Normal prostate and some malignant prostate cancer (PrCa) cell lines undergo acinar differentiation and form spheroids in three-dimensional (3-D) organotypic culture. Acini formed by PC-3 and PC-3M, less pronounced also in other PrCa cell lines, spontaneously undergo an invasive switch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service