Skip to Content
Merck
All Photos(4)

Key Documents

HPA003711

Sigma-Aldrich

Anti-NPNT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Nephronectin precursor, Anti-Preosteoblast EGF-like repeat protein with MAM domain, Anti-Protein EGFL6-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

RQCVNTFGSYICKCHKGFDLMYIGGKYQCHDIDECSLGQYQCSSFARCYNVRGSYKCKCKEGYQGDGLTCVYIPKVMIEPSGPIHVPKGNGTILKGDTGNNNWIPDVGSTWWPPKTPYIPPIITNRPTSKPTTRPTPK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPNT(255743)

General description

Nephronectin (NPNT) is an extracellular matrix protein consisting of five EGF-like repeats, a mucin region containing a RGD sequence, and a COOH-terminal MAM domain. In humans, it is located at the chromosomal position 4q25. It has two transcript variants (i.e. NPNTa and NPNTb). It is expressed in the embryonic (ear, eye, heart, lung and kidney) and adult tissues (in lung, kidney, brain, uterus, placenta, thyroid gland and blood vessels).

Immunogen

Nephronectin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Nephronectin (NPNT) is involved in the embryonic development and maintenance of various adult tissues. It functions in the differentiation of osteoblast via epidermal growth factor-like repeats. It also participates in the development of embryonic kidney with integrin α8β1.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85230

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shireen Kahai et al.
FEBS letters, 584(1), 233-238 (2009-12-01)
We found that nephronectin was significantly down-regulated by TGF-beta1. To determine the function of nephronectin in osteogenesis, we generated various constructs to produce stable MC3T3-E1 cell lines, expressing and secreting nephronectin protein, including full-length (Npnt), lacking EGF-like repeats (Np-MAM), and
Tonje S Steigedal et al.
Neoplasia (New York, N.Y.), 20(4), 387-400 (2018-03-15)
Most cancer patients with solid tumors who succumb to their illness die of metastatic disease. While early detection and improved treatment have led to reduced mortality, even for those with metastatic cancer, some patients still respond poorly to treatment. Understanding
Tobias Åkerström et al.
Endocrine-related cancer, 22(5), 735-744 (2015-08-20)
Aldosterone-producing adenomas (APAs) are found in 1.5-3.0% of hypertensive patients in primary care and can be cured by surgery. Elucidation of genetic events may improve our understanding of these tumors and ultimately improve patient care. Approximately 40% of APAs harbor
Jeffrey T-J Huang et al.
International journal of molecular medicine, 15(4), 719-724 (2005-03-09)
We describe the identification and characterization of a human epidermal growth factor (EGF)-like gene, nephronectin (NPNT), which encodes predicted proteins most homologous to the mouse nephronectin (Npnt). Human NPNT is located at the chromosomal position 4q25. Two transcript variants, NPNTa
R Brandenberger et al.
The Journal of cell biology, 154(2), 447-458 (2001-07-27)
The epithelial-mesenchymal interactions required for kidney organogenesis are disrupted in mice lacking the integrin alpha8beta1. None of this integrin's known ligands, however, appears to account for this phenotype. To identify a more relevant ligand, a soluble integrin alpha8beta1 heterodimer fused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service