Skip to Content
Merck
All Photos(7)

Key Documents

HPA003097

Sigma-Aldrich

Anti-NF2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Merlin antibody produced in rabbit, Anti-Moesin-ezrin-radixin-like protein antibody produced in rabbit, Anti-Neurofibromin-2 antibody produced in rabbit, Anti-Schwannomerlin antibody produced in rabbit, Anti-Schwannomin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ERRAKQKLLEIATKPTYPPMNPIPAPLPPDIPSFNLIGDSLSFDFKDTDMKRLSMEIEKEKVEYMEKSKHLQEQLNELKTEIEALKLKERETALDILHNENSDRGGSSKHNTIKKLTLQSAKSRVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NF2(4771)

General description

NF2 is a tumor suppressor present in the nucleus and membranes of cells. NF2 modulates mitotic inhibition, and also regulates the adhesion and polarity of epithelial cells. Additionally, this protein facilitates contact inhibition . Anti-NF2 antibody is specific for NF2 in humans.

Immunogen

Merlin recombinant protein epitope signature tag (PrEST)

Application

Anti-NF2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-NF2 antibody is also suitable for use in indirect immunofluorescence assays.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73592

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Xue et al.
American journal of translational research, 13(8), 8787-8803 (2021-09-21)
A stable, human sporadic vestibular schwannoma cell line is not currently available. By using a lentivirus-mediated transfection from a 41-year-old sporadic vestibular schwannoma patient, primary schwannoma cells were obtained, cultured and immortalized using the hHERT gene. The NF2 gene of
Lu Zhou et al.
FEBS letters, 586(10), 1403-1408 (2012-05-19)
Recent evidence suggests that the neurofibromatosis type 2 (NF2) gene encoded protein merlin suppresses mitogenic signalling not only at the cell membrane but also in the nucleus. At the membrane, merlin inhibits signalling by integrins and tyrosine receptor kinases (RTKs)
Wei Li et al.
EMBO reports, 13(3), 204-215 (2012-04-07)
Inhibition of proliferation by cell-to-cell contact is essential for tissue organization, and its disruption contributes to tumorigenesis. The FERM domain protein Merlin, encoded by the NF2 tumour suppressor gene, is an important mediator of contact inhibition. Merlin was thought to
Alexander Laure et al.
Cancers, 15(10) (2023-06-22)
Cell lines are extensively used to study cancer biology. However, the use of highly passaged commercial cell lines has to be questioned, as they do not closely resemble the originating tumor. To understand the reliability of preclinical models for Malignant
Kana T Furukawa et al.
Cell reports, 20(6), 1435-1447 (2017-08-10)
Circumferential actin belts underlying the adherens junctions of columnar epithelial cell monolayers control intercellular surface tension and cell shape to maintain tissue integrity. Yes-associated protein (YAP) and its paralog TAZ are proliferation-activating transcriptional coactivators that shuttle between the nucleus and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service