Skip to Content
Merck
All Photos(5)

Key Documents

HPA002834

Sigma-Aldrich

Anti-Anti-PTGS1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-COX1, Anti-PGHS-1, Anti-PTGHS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MPDSFKVGSQEYSYEQFLFNTSMLVDYGVEALVDAFSRQIAGRIGGGRNMDHHILHVAVDVIRESREMRLQPFNEYRKRFGMKPYTSFQELVGEKEMAAELEELYGDIDALEFYPGLLLEKCHPNSIFGESMIEIGAPFSLKGLLGN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGS1(5742)

Immunogen

Prostaglandin G/H synthase 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cytochrome c oxidase (COX) is the terminal enzyme of the respiratory chain having subunits originating both from the mitochondrial and nuclear genome. Deficiency of this gene causes a MELAS-like syndrome. Its biogenesis defect does not involve a primary alteration in the expression of the COX subunits. Its expression is translationally regulated as in most bona fide COX assembly mutants. It is found that assembled ATPase and its proton pumping function are essential for COX biogenesis in yeast and mammalian cells through a mechanism independent of Cox1p synthesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83040

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E W Y Tam et al.
Neuropediatrics, 39(6), 328-334 (2009-07-02)
Cytochrome c oxidase (COX) is the terminal enzyme of the respiratory chain, with subunits originating both from the mitochondrial and nuclear genome. An eleven-year-old female presented initially with a seizure followed two months later with tonic-clonic seizures, weakness and aphasia.
Daniel Bergemalm et al.
Haematologica, 106(11), 2947-2959 (2020-10-16)
In X-linked thrombocytopenia with thalassemia (XLTT; OMIM 314050), caused by the mutation p.R216Q in exon 4 of the GATA1 gene, male hemizygous patients display macrothrombocytopenia, bleeding diathesis and a β-thalassemia trait. Herein, we describe findings in two unrelated Swedish XLTT
Wanting Shao et al.
Journal of translational medicine, 18(1), 94-94 (2020-02-23)
The aim of this study was to investigate the expression of the nuclear receptor PPARγ, together with that of the cyclooxygenases Cox-1 and Cox-2, in breast cancer (BC) tissues and to correlate the data with several clinicobiological parameters including patient
Ileana C Soto et al.
Biochimica et biophysica acta, 1793(11), 1776-1786 (2009-09-09)
The mitochondrial F(1)F(0)-ATP synthase or ATPase is a key enzyme for aerobic energy production in eukaryotic cells. Mutations in ATPase structural and assembly genes are the primary cause of severe human encephalomyopathies, frequently associated with a pleiotropic decrease in cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service