Skip to Content
Merck
All Photos(5)

Key Documents

HPA002657

Sigma-Aldrich

Anti-CPM antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Carboxypeptidase M precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPM(1368)

Related Categories

General description

CPM (carboxypeptidase M) is an extracellular glycosylphosphatidylinositol (GPI) anchored membrane glycoprotein. It belongs to the regulatory or CPN/E subfamily of zinc metallocarboxypeptidases. It consists of a catalytic domain at the N-terminal end, conically shaped β-sandwich C-terminal domain characteristic of the CPN/E family and C-terminal extension to which the GPI (glycosylphosphatidylinositol) membrane-anchor is post-translationally attached.

Immunogen

Carboxypeptidase M precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CPM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CPM (carboxypeptidase M) specifically acts in the removal of C-terminal basic residues from peptides and proteins. It may regulate the peptide hormone and growth factor activity at the cell surface, and the membrane-localized degradation of extracellular proteins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85162

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine J Denis et al.
Clinica chimica acta; international journal of clinical chemistry, 415, 306-316 (2012-11-28)
This review covers carboxypeptidase M (CPM) research that appeared in the literature since 2009. The focus is on aspects that are new or interesting from a clinical perspective. Available research tools are discussed as well as their pitfalls and limitations.
Fulong Tan et al.
The Biochemical journal, 370(Pt 2), 567-578 (2002-11-30)
Human carboxypeptidase (CP) M was expressed in baculovirus-infected insect cells in a glycosylphosphatidylinositol-anchored form, whereas a truncated form, lacking the putative signal sequence for glycosylphosphatidylinositol anchoring, was secreted at high levels into the medium. Both forms had lower molecular masses
David Reverter et al.
Journal of molecular biology, 338(2), 257-269 (2004-04-07)
Carboxypeptidase M (CPM), an extracellular glycosylphosphatidyl-inositol(GPI)-anchored membrane glycoprotein belonging to the CPN/E subfamily of "regulatory" metallo-carboxypeptidases, specifically removes C-terminal basic residues from peptides and proteins. Due to its wide distribution in human tissues, CPM is believed to play important roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service