Skip to Content
Merck
All Photos(6)

Key Documents

HPA001925

Sigma-Aldrich

Anti-TAGLN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SM22-alpha homolog, Anti-Transgelin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RGPAYGLSREVQQKIEKQYDADLEQILIQWITTQCRKDVGRPQPGRENFQNWLKDGTVLCELINALYPEGQAPVKKIQASTMAFKQMEQISQFLQAAERYGINTTDIFQTVDLWEGKNMACVQRTLMNLGGLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAGLN2(8407)

General description

TAGLN2 (transgelin 2) was first identified as a marker of differentiated smooth muscle. It is expressed in the adult smooth muscle cells and embryonic skeletal and cardiac tissues. Its function has not yet clear but it has been reported that TAGLN2 may plays a role in oncogenic function. Tumor suppressive microRNA (miRNA) gene regulates the TAGLN2 activity in head and neck squamous cell carcinoma (HNSCC).

Immunogen

Transgelin-2 recombinant protein epitope signature tag (PrEST)

Application

Anti-TAGLN2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86390

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Stanier et al.
Genomics, 51(1), 144-147 (1998-08-07)
SM22alpha (TAGLN) is one of the earliest markers of differentiated smooth muscle, being expressed exclusively in the smooth muscle cells of adult tissues and transiently in embryonic skeletal and cardiac tissues. We have identified and mapped the mouse Tagln gene
H Yoshino et al.
British journal of cancer, 104(5), 808-818 (2011-02-10)
On the base of the microRNA (miRNA) expression signature of bladder cancer (BC), we found that miR-1 and miR-133a were significantly downregulated in BC. In this study, we focussed on the functional significance of miR-1 and miR-133a in BC cell
Nijiro Nohata et al.
Oncotarget, 2(1-2), 29-42 (2011-03-08)
Based on the microRNA (miRNA) expression signatures of hypopharyngeal and esophageal squamous cell carcinoma, we found that miR-1 was significantly down-regulated in cancer cells. In this study, we investigated the functional significance of miR-1 in head and neck squamous cell
Asa Bolander et al.
Cancer genomics & proteomics, 5(6), 293-300 (2009-03-17)
Patients with metastazing malignant melanoma have a poor outcome and determination of thickness of the primary tumor remains as the most important prognostic predictor. The aim of this study was to use an antibody-based proteomics strategy to search for new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service