Skip to Content
Merck
All Photos(6)

Key Documents

HPA001912

Sigma-Aldrich

Anti-TFAP4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Activating enhancer-binding protein 4 antibody produced in rabbit, Anti-Transcription factor AP-4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFAP4(7023)

General description

Transcription factor, TFAP4 (transcription factor AP-4), is a helix-loop-helix (HLH) protein with multiple distinct protein-protein interfaces. Its protein dimerization motif contains leucine repeat elements LR1 and LR2. The HLH domain is responsible for site-specific DNA binding. LR1 and LR2 are also involved with DNA recognition and binding mechanisms along with the principal HLH domain.

Immunogen

Transcription factor AP-4 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Immunohistochemistry (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84739

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung Kyung Choi et al.
Oncology reports, 35(2), 1186-1194 (2015-11-18)
Bromodomain and extra-terminal domain (BET) family proteins are representative epigenetic modulators that read acetylated lysine residues and transfer cellular signals. Recently, the BET protein inhibitor JQ1 was developed and has been extensively studied in many cancer cell types. We demonstrated
Chengyuan Xue et al.
Oncotarget, 7(34), 54937-54951 (2016-07-28)
Amplification of the MYCN oncogene, a member of the MYC family of transcriptional regulators, is one of the most powerful prognostic markers identified for poor outcome in neuroblastoma, the most common extracranial solid cancer in childhood. While MYCN has been
Y F Hu et al.
Genes & development, 4(10), 1741-1752 (1990-10-01)
Enhancer binding protein AP-4 is a transcription factor that activates both viral and cellular genes by binding to the symmetrical DNA sequence, CAGCTG. Here, we report the molecular cloning and characterization of human AP-4 cDNAs. The deduced amino acid sequence
Andrea J Savio et al.
Epigenetics, 12(6), 441-448 (2017-03-18)
The MLH1 promoter polymorphism rs1800734 is associated with MLH1 CpG island hypermethylation and expression loss in colorectal cancer (CRC). Conversely, variant rs1800734 is associated with MLH1 shore, but not island, hypomethylation in peripheral blood mononuclear cell DNA. To explore these
Weiwei Wang et al.
Bioengineered, 13(1), 1626-1636 (2022-01-11)
Hepatocellular carcinoma (HCC) is a highly vascularized solid tumor with a fast growth rate. According to bioinformatics analysis, CC chemokine ligand 23 (CCL23) has clinical significance for survival and prognosis in HCC. The online databases TCGA and CCLE were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service