Skip to Content
Merck
All Photos(4)

Key Documents

HPA001873

Sigma-Aldrich

Anti-ACTN4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-Actinin-4 antibody produced in rabbit, Anti-F-actin cross-linking protein antibody produced in rabbit, Anti-Non-muscle α-actinin 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

MFIVHTIEEIEGLISAHDQFKSTLPDADREREAILAIHKEAQRIAESNHIKLSGSNPYTTVTPQIINSKWEKVQQLVPKRDHALLEEQSKQQSNEHLRRQFASQANVVGPWIQTKMEEIGRISIEMNGTLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTN4(81)

Immunogen

α-Actinin-4 recombinant protein epitope signature tag (PrEST)

Application

Anti-ACTN4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

α actinins, actin-filament crosslinking proteins, belong to the spectrin family. The two non-muscle isoforms α-actinin-1 and α-actinin-4 are expressed in podocytes. At the C terminus of ACTN4 (actinin α 4) contains a PDZ (PSD95, Dlg1 and zo-1) domain-binding motif that binds to membrane-associated guanylate kinase inverted-1 (MAGI-1) in actin cytoskeleton dynamics. It is associated with chromatin remodeling and other nuclear events. Mutation in ACTN4 gene causes nephrotic syndrome, focal and segmental glomerulosclerosis (FSGS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85161

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark T Poch et al.
Toxicology and applied pharmacology, 199(3), 239-250 (2004-09-15)
A negative regulatory element (NRE1; position -794 to -774) was previously identified that mediates the downregulation of CYP1A1, including partial suppression of Ah receptor-dependent induction. The CCAAT-box binding protein, nuclear factor-Y (NF-Y), is a component of one of two protein
O Ichii et al.
Histology and histopathology, 26(10), 1243-1255 (2011-08-27)
Podocytes cover the glomerulus and their adjacent foot processes form a principal barrier called the slit diaphragm. Podocyte dysfunctions, including podocyte loss and slit diaphragm disruptions, induce chronic kidney diseases (CKD). In this study, we analyzed the correlations between podocyte
Sharon P Andreoli
Advances in renal replacement therapy, 11(1), 105-109 (2004-01-20)
Idiopathic focal segmental glomerulosclerosis (FSGS) is a common cause of nephrotic syndrome in pediatric and adult patients. Most children with FSGS do not respond to any form of therapy and progress to end-stage renal disease (ESRD). FSGS reoccurs in the
M J F Broderick et al.
Advances in protein chemistry, 70, 203-246 (2005-04-20)
Spectrin family proteins represent an important group of actin-bundling and membrane-anchoring proteins found in diverse structures from yeast to man. Arising from a common ancestral alpha-actinin gene through duplications and rearrangements, the family has increased to include the spectrins and
Magdalena Misiura et al.
The American journal of the medical sciences, 359(3), 147-155 (2020-01-01)
Fascin-1 and actinin-4 are involved in key processes of tumor cell adhesion, migration and metastasis. Actinin-4 plays an important role in promotion of cell proliferation, whereas fascin-1 regulates cellular motility. Its over-expression leads to the loss of cell adhesion and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service