Skip to Content
Merck
All Photos(5)

Key Documents

HPA001119

Sigma-Aldrich

Anti-YY1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Delta transcription factor, Anti-NF-E1, Anti-Transcriptional repressor protein YY1, Anti-YY-1, Anti-Yin and yang 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DLSDPKQLAEFARMKPRKIKEDDAPRTIACPHKGCTKMFRDNSAMRKHLHTHGPRVHVCAECGKAFVESSKLKRHQLVHTGEKPFQCTFEGCGKRFSLDFNLRTHVRIHTGDRPYVCPFDGCNKKFAQSTNLKSHILTH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YY1(7528)

General description

The gene YY1 (Yin Yang 1) encodes a protein belonging to the GLI-Krüppel gene family. It is a zinc finger protein that is ubiquitously expressed. The gene is mapped to human chromosome 14q32.

Immunogen

Transcriptional repressor protein YY1 recombinant protein epitope signature tag (PrEST)

Application

Anti-YY1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

YY1 (Yin Yang 1) is a transcriptional repressor protein encoded by the YY1 gene in humans. It regulates diverse biological processes and may have an important role in carcinogenesis. Its expression is found to be upregulated in gastric cancer cell lines and primary gastric cancers. YY1 is found to contribute to carcinogenesis in gastric cancer. This DNA-binding protein is found to regulate the activity of the c-fos promoter.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70355

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha Kaufhold et al.
Journal of experimental & clinical cancer research : CR, 35, 84-84 (2016-05-27)
The transcription factor Yin Yang 1 (YY1) is frequently overexpressed in cancerous tissues compared to normal tissues and has regulatory roles in cell proliferation, cell viability, epithelial-mesenchymal transition, metastasis and drug/immune resistance. YY1 shares many properties with cancer stem cells
S Natesan et al.
Genes & development, 7(12B), 2497-2509 (1993-12-01)
The assembly of multicomponent complexes at promoters, enhancers, and silencers likely entails perturbations in the path of the DNA helix. We present evidence that YY1, a ubiquitously expressed DNA-binding protein, regulates the activity of the c-fos promoter primarily through an
Y Shi et al.
Cell, 67(2), 377-388 (1991-10-18)
A sequence within the transcription control region of the adeno-associated virus P5 promoter has been shown to mediate transcriptional activation by the adenovirus E1A protein. We report here that this same element mediates transcriptional repression in the absence of E1A.
Valentina Bevelacqua et al.
Oncotarget, 3(8), 882-892 (2012-08-30)
NECL-5 is involved in regulating cell-cell junctions, in cooperation with cadherins, integrins and platelet-derived growth factor receptor, that are essential for intercellular communication. Its role in malignant transformation was previously described. It has been reported that transformation of melanocytes is
Dharmaraj Chinnappan et al.
International journal of oncology, 34(5), 1417-1423 (2009-04-11)
Over-expression of the multifunctional zinc-finger transcription factor Yin Yang 1 (YY1) has been associated with cellular proliferation and resistance to apoptotic stimuli. In this study, we report that YY1 was uniformly highly over-expressed in a wide range of human cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service