Skip to Content
Merck
All Photos(2)

Key Documents

H7154

Sigma-Aldrich

N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid

BioXtra, ≥98%

Synonym(s):

N-Carboxymethyl-N′-(2-hydroxyethyl)-N,N′-ethylenediglycine, HEDTA, HEEDTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2N(CH2CO2H)CH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
278.26
Beilstein:
1804795
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

product line

BioXtra

Quality Level

Assay

≥98%

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤1%

mp

212 °C (dec.) (lit.)

solubility

3  M NaOH: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤1%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.5%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OCCN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H18N2O7/c13-4-3-11(5-8(14)15)1-2-12(6-9(16)17)7-10(18)19/h13H,1-7H2,(H,14,15)(H,16,17)(H,18,19)

InChI key

URDCARMUOSMFFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid (HEDTA) is a chelating agent that is used to form complexes with metal ions such as iron(II), zinc(II), manganese(II) and uranium(VI) ions. It can also be used to prepare fluorescent carbon dots (CDs).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sunset yellow in soft drinks based on fluorescence quenching of carbon dots.
Yuan, Yusheng et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 167(22), 106-110 (2016)
Complexation of Uranium (VI) with N-(2-Hydroxyethyl) ethylenediamine-N, N?, N?-triacetic Acid in Aqueous Solution: Thermodynamic Studies and Coordination Analyses.
Li, Xingliang et al.
Inorganic Chemistry, 204(11), 1391-1397 (2018)
A novel manganese complex effective as superoxide anion scavenger and therapeutic agent against cell and tissue oxidative injury.
Failli, Paola et al.
Journal of Medicinal Chemistry, 52(22), 7273-7283 (2009)
Supramolecular branching and crosslinking of terpyridine-modified copolymers: complexation and decomplexation studies in diluted solution.
Hofmeier, Harald and Schubert, Ulrich S
Macromolecular Chemistry and Physics, 204(11), 1391-1397 (2003)
Swaran J S Flora et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 134(3), 319-328 (2003-03-20)
Aluminum, a known neurotoxic substance, has been suggested as a contributing factor in the pathogenesis of Alzheimer's disease. Therapeutic efficacy of combined administration of citric acid (CA) and N-(2-hydroxyethyl) ethylenediaminetriacetic acid (HEDTA) was evaluated in decreasing blood and brain aluminum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service