Skip to Content
Merck
All Photos(1)

Key Documents

H7041

Sigma-Aldrich

IL-2 human

≥98% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-2, T-cell growth factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

form

lyophilized powder

potency

≤5.0 ng/mL ED50

quality

endotoxin tested

mol wt

monomer 15 kDa (glycosylated)

packaging

pkg of 5X10 μg
pkg of 10 μg
pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL2(3558)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-2 (IL-2) is an α-helical cytokine belonging to T-helper cell type 1. It has a molecular weight of 15kDa. The IL2 gene is mapped to human chromosome 4q27. IL-2 is originally called as T cell growth factor.

Application

Interleukin-2 human has been used to culture peripheral blood mononuclear cells (PBMC). It has been used to activate effector cell (PBMC) and to study antibody-dependent cell-mediated cytotoxicity.

Biochem/physiol Actions

Interleukin-2 (IL-2) is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lympho­kine promotes long term growth of acti­vated T cells and related cell types. IL-2 plays a role in the activation and proliferation of NK cells, induces γ-inter­feron and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.
Interleukin-2 (IL-2) is essential for proliferation of natural killer (NK) cells, lymphocytes and macrophages. It plays a key role in the immune system functions such as NK cell-stimulated immune responses. IL-2 also mediates the formation and essential functions of regulatory T cells. It is implicated in autoimmune disease. IL-2 is associated with apoptosis and thus, possesses antitumor response. IL-2 mediates CD41+ T-cells differentiation into T-helper cell 1 and 2 effector subsets, preventing T-helper 17 differentiation.

Preparation Note

Human Interleukin-2 (IL-2) expressed in human HEK 293 cells is a glycosylated monomer with a molecular mass of 15 kDa. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent stimulation of the proliferation of mouse CTLL-2 cells (mouse cytotoxic T cell line).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-glycan engineering of a plant-produced anti-CD20-hIL-2 immunocytokine significantly enhances its effector functions
Marusic C, et al,
Biotechnology and Bioengineering, 115(3), 565-576 (2018)
Interleukin 2, 37-37 (2012)
Functional incompatibility between the generic NF-? motif and a subtype-specific Sp1III element drives the formation of HIV-1 subtype C viral promoter
Verma A, et al.
Journal of Virology, JVI-00308 (2016)
Association of- 330 interleukin-2 gene polymorphism with oral cancer
Singh PK, et al.
The Indian Journal of Medical Research, 146(6), 730-730 (2017)
Association of Interleukin-2-330T/G and Interleukin-10-1082A/G Genetic Polymorphisms with B-Cell Non-Hodgkin Lymphoma in a Cohort of Egyptians
Rahman HAA, et al.
Turkish journal of haematology : official journal of Turkish Society of Haematology, 35(2), 99-99 (2018)

Articles

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service