Skip to Content
Merck
All Photos(2)

Key Documents

H6268

Sigma-Aldrich

Hide–Remazol Brilliant Blue R

protease substrate, powder

Synonym(s):

Hide powder azure, Remazol Brilliant Blue R–Hide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5629635
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Hide–Remazol Brilliant Blue R, protease substrate

Quality Level

form

powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Hide−Remazol Brilliant Blue R has been used:
  • to study the substrate specificity of a purified collagenase preparation from Clostridium histolyticum[1]
  • to mix with cytotoxic samples for hide powder azure protease activity assay[2]
  • as a high molecular weight substrate to test the inhibition of trypsin by the purified recombinant rat bikunin, α1-m/bikunin precursor, and α1-m[3]
  • as a substrate to obtain the inhibition of snake venom protease activity by the addition of exogenous citrate[4]

Other Notes

Hide powder covalently linked to Remazol Brilliant Blue R

Substrates

Chromogenic substrate for trypsin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Citrate inhibition of snake venom proteases
Odell G V, et al.
Toxicon, 36(12), 1801-1806 (1998)
Expression of a functional proteinase inhibitor capable of accepting xylose: bikunin
Falkenberg C, et al.
Archives of Biochemistry and Biophysics, 387(1), 99-106 (2001)
Soluble, dye-labelled substrates for a micro-plate assay of proteinase activity
Wolf G A and Wirth S J
Journal of Microbiological Methods, 25(3), 337-342 (1996)
C Kooi et al.
Journal of bacteriology, 187(13), 4421-4429 (2005-06-22)
Burkholderia cenocepacia ZmpA is expressed as a preproenzyme typical of thermolysin-like proteases such as Pseudomonas aeruginosa LasB and Bacillus thermoproteolyticus thermolysin. The zmpA gene was expressed using the pPRO-EXHTa His(6) tag expression system, which incorporates a six-His tag at the
Boophilus microplus: multiple proteolytic activities in the midgut.
J Mendiola et al.
Experimental parasitology, 82(1), 27-33 (1996-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service