Skip to Content
Merck
All Photos(1)

Key Documents

H4914

Sigma-Aldrich

Honokiol

≥98% (HPLC), powder

Synonym(s):

5,3′-Diallyl-2,4′-dihydroxybiphenyl, NSC 293100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

DMSO: 36 mg/mL

antibiotic activity spectrum

Gram-positive bacteria
neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1CC=C)-c2cc(CC=C)ccc2O

InChI

1S/C18H18O2/c1-3-5-13-7-9-18(20)16(11-13)14-8-10-17(19)15(12-14)6-4-2/h3-4,7-12,19-20H,1-2,5-6H2

InChI key

FVYXIJYOAGAUQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Honokiol exhibits analgesic, antithrombotic, antispasmodic, antidepressant, antimicrobial and neuroprotective effects. It regulates gamma-aminobutyric acid (GABA) A receptors. Honokiol is associated with an increased risk of bleeding. It prevents platelet aggregation, protects myocardium from ischemic damage and inhibits ventricular arrhythmia.

Application

Honokiol has been used:
  • to study its effects on plasmid hSirt3102-399 deacetylation activity
  • as an antioxidant to study its cytoprotective role in human ovarian cancer cells (SKOV-3) and Chinese hamster ovary cells (CHOK1)
  • to explore its effects on oxidative stress and mitochondrial dysfunction via a sirt3-dependent manner
  • for intracerebroventricular (ICV) cannulation

Biochem/physiol Actions

Honokiol is a natural biphenyl neolignan from magnolia extract. It is antiangiogenic, antitumor, and anxiolytic.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shing Hwa Liu et al.
PloS one, 7(8), e43711-e43711 (2012-09-01)
Honokiol, a small-molecular weight natural product, has previously been reported to activate apoptosis and inhibit gastric tumorigenesis. Whether honokiol inhibits the angiogenesis and metastasis of gastric cancer cells remains unknown. We tested the effects of honokiol on angiogenic activity and
Honokiol, a constituent of oriental medicinal herb magnolia officinalis, inhibits growth of PC-3 xenografts in vivo in association with apoptosis induction
Hahm ER, et al.
Clinical Cancer Research, 14(4), 1248-1257 (2008)
Mild Impairment of Mitochondrial OXPHOS Promotes Fatty Acid Utilization in POMC Neurons and Improves Glucose Homeostasis in Obesity
Timper K, et al.
Testing, 25(2), 383-397 (2018)
Sirt3 ameliorates oxidative stress and mitochondrial dysfunction after intracerebral hemorrhage in diabetic rats
Zheng J, et al.
Frontiers in Neuroscience, 12, 526376-526376 (2018)
The role of phospholipase D in modulating the MTOR signaling pathway in polycystic kidney disease
Liu Y, et al.
PLoS ONE, 8(8), e73173-e73173 (2013)

Articles

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service