Skip to Content
Merck
All Photos(1)

Key Documents

H4149

Sigma-Aldrich

Monoclonal Anti-Heat Shock Protein 60 antibody produced in mouse

clone LK1, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LK1, monoclonal

mol wt

antigen 60 kDa

contains

15 mM sodium azide

species reactivity

rat, human, chicken

technique(s)

electron microscopy: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1:400 using cultured human foreskin fibroblast extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPD1(3329)
rat ... Hspd1(63868)

General description

Heat shock (stress) proteins (HSP) are produced by prokaryotic and eukaryotic cells, and are among the most conserved molecules in phylogeny. HSP-60 is a member of HSP family with a uniquely high level of sequence conservation.
Monoclonal Anti-Heat Shock Protein 60 (mouse IgG1 isotype) is derived from the LK1 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with recombinant human heat diseases.

Specificity

The antibody recognizes an epitope located between amino acid residues 383-447 of the human HSP 60. It is not cross-reactive with the bacterial counterpart (E. coli), or with helminth and spinach HSP60. The antibody reacts with constitutive and inducible HSP60. It demonstrates a raised level of staining in immunohistochemistry of synovial membranes taken from patients with juvenile chronic arthritis.

Immunogen

recombinant human heat shock protein 60 (HSP60)

Application

Monoclonal Anti-Heat Shock Protein 60 has been used in the following techniques:
  • Enzyme linked immunosorbent assay (ELISA)
  • Immunoblotting
  • Immunocytochemistry
  • Immunohistochemistry
  • Immunoelectron microscopy
  • Double immunofluorescence

Biochem/physiol Actions

A wide variety of environmental perturbations, such as a sudden increase in temperature, induce cells to rapidly synthesize a group of polypeptides known as the heat shock (stress) proteins. HSP-60 is considered to be a potential antigen in a number of autoimmune diseases. Abnormal immune reactivity involving shock protein 60 (HSP60) has also been implicated in the pathogenesis of schizophrenia. This monoclonal antibody enables the differentiation between the HSP60 of mammalian and bacterial origin.
Abnormal immune reactivity involving HSP60 has also been implicated in the pathogenesis of schizophrenia.

Physical form

Monoclonal Anti-Heat Shock Protein 60 is provided as ascites fluid with 0.1% sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Patterson et al.
Cell death and differentiation, 7(2), 137-144 (2000-03-14)
Mitochondrial membrane permeabilization is a rate-limiting step of cell death. This process is, at least in part, mediated by opening of the permeability transition pore complex (PTPC) Several soluble proteins from the mitochondrial intermembrane space and matrix are involved in
Identification of antibodies to heat shock proteins 90 kDa and 70 kDa in patients with schizophrenia
Kim JJ, et al.
Schizophrenia Research, 52(1-2), 127-135 (2001)
Heat shock protein 60 levels in tissue and circulating exosomes in human large bowel cancer before and after ablative surgery
Campanella C, et al.
Cancer, 121(18), 3230-3239 (2015)
Elevated blood Hsp60, its structural similarities and cross-reactivity with thyroid molecules, and its presence on the plasma membrane of oncocytes point to the chaperonin as an immunopathogenic factor in Hashimoto's thyroiditis
Gammazza A, et al.
Cell Stress & Chaperones, 19(3), 343-353 (2014)
Hsp60 and Hsp10 increase in colon mucosa of Crohn?s disease and ulcerative colitis
Rodolico V, et al.
Cell Stress & Chaperones, 15(6), 877-884 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service